Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00520959_g1
          See other LRRC14 GE Assays ›
          Gene Symbol:
          Lrrc14
          Gene Name:
          leucine rich repeat containing 14
          Gene Aliases:
          E130306I01Rik, mKIAA0014
          Chromosome Location:
          Chr.15: 76710623 - 76715097 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00520959_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lrrc14
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_145471.2 NP_663446.1 5 - 6 1269 65
          GenBank mRNA AK036278.1 - 3 - 4 1142 65
          AK047414.1 - 3 - 4 814 65
          AK053747.1 - 1379 65
          AK132158.1 - 3 - 4 1833 65
          AK149655.1 - 3 - 4 1232 65
          AK159836.1 - 3 - 4 1212 65
          AK172871.1 - 3 - 4 1808 65
          AY239225.1 - 3 - 4 1503 65
          AY239226.1 - 5 - 6 1185 65
          BC022126.1 - 5 - 6 1302 5097371 65
          BC033482.1 - 2 - 3 1236 65
          BC138217.1 - 3 - 4 1443 8861236 65

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1500000607.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Lrrc14

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30350131 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30350133 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P639101 AK172871.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P639101 BC033482.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P639101 BC138217.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P639101 BC145203.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P639101 AY239226.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P639101 AK172871.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P639101 BC033482.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P639101 BC138217.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P639101 BC145203.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P639101 AY239226.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 462798 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 624518 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 99961 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P639101 AK172871.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P639101 BC033482.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P639101 BC138217.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P639101 BC145203.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P639101 AY239226.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P639101 AK172871.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P639101 BC033482.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P639101 BC138217.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P639101 BC145203.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P639101 AY239226.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30350137 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30350162 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30350158 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30350134 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30350139 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30350147 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30350151 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30350149 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30350133 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30350159 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30350157 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30350153 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30350152 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30350140 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30350150 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30350145 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30350155 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30350142 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30350131 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30350144 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30350161 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08423200 - -
          Whole Mouse Genome G4122A A_52_P639101 AK172871.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P639101 BC033482.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P639101 BC138217.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P639101 BC145203.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P639101 AY239226.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P639101 AK172871.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P639101 BC033482.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P639101 BC138217.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P639101 BC145203.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P639101 AY239226.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P126684 AK053747.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P126684 AK042532.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P104779 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P110429 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12258122 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12203772 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12203778 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30350131 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30350133 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350137 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350150 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350140 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350134 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350194 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350186 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350149 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350153 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350142 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350185 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350192 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350155 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350187 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350213 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350072 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350215 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350195 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350205 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350220 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350182 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350157 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350223 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350224 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350217 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350162 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350159 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350145 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350128 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350161 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350190 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350202 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350189 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350212 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350152 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350177 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350147 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350219 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350211 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350208 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350183 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350133 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350210 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350199 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350131 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30350155 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12203778 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30350150 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30350147 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12119740 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30350158 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30350145 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30350134 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30350162 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30350141 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30350152 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12351246 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30350153 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12203774 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30350149 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30350159 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30350140 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30350157 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12258122 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30350151 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30350161 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10425007 - -
          Affymetrix Mouse Gene 1.0 ST Array 10425002 - -
          Affymetrix Mouse Gene 1.0 ST Array 10425009 - -
          Affymetrix Mouse Gene 1.0 ST Array 10425006 - -
          Affymetrix Mouse Gene 1.0 ST Array 10425003 - -
          Affymetrix Mouse Gene 1.0 ST Array 10425010 - -
          Affymetrix Mouse Gene 1.0 ST Array 10425001 - -
          Affymetrix Mouse Gene 1.0 ST Array 10425005 - -
          Affymetrix Mouse Gene 1.0 ST Array 10425008 - -
          Affymetrix Mouse Gene 1.0 ST Array 10425004 - -
          Affymetrix HT MG-430 PM Array Plate 1424419_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10425000 - -
          Mouse Genome 430 2.0 Array 1445666_PM_at - -
          Mouse Genome 430 2.0 Array 1440470_PM_at - -
          Mouse Genome 430 2.0 Array 1424419_PM_at - -
          Mouse Genome 430 2.0 Array 1445666_at - -
          Mouse Genome 430 2.0 Array 1440470_at - -
          Mouse Genome 430 2.0 Array 1424419_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 223664_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 223664_at - -
          Affymetrix Mouse Clariom S Array PSR1500004462.mm.1 NM_145471.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500004462.mm.1 NM_145471.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1500004462.mm.1 TC1500000607.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500004460.mm.1 NM_145471.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500004460.mm.1 NM_145471.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1500004460.mm.1 TC1500000607.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500004463.mm.1 NM_145471.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500004463.mm.1 NM_145471.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1500004463.mm.1 TC1500000607.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500004458.mm.1 NM_145471.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500004458.mm.1 NM_145471.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1500004458.mm.1 TC1500000607.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500004461.mm.1 NM_145471.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500004461.mm.1 NM_145471.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1500004461.mm.1 TC1500000607.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500004465.mm.1 NM_145471.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500004465.mm.1 NM_145471.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1500004465.mm.1 TC1500000607.mm.1 MTA TC
          Mouse Expression Set 430 1424419_PM_at - -
          Mouse Expression Set 430 1424419_at - -
          Mouse Genome 430A 2.0 Array 1424419_PM_at - -
          Mouse Genome 430A 2.0 Array 1424419_at - -
          Affymetrix HT Mouse Genome 430A Array 1424419_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1424419_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 5080465 - -
          Illumina mouse-6 v1.0 expression beadchip 5080465 - -
          Illumina mouse-6 v1.0 expression beadchip 6650440 - -
          Illumina mouse-6 v1.0 expression beadchip 106100494 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005080465 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006650440 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2758168 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2626606 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2626606 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2758168 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2457925 - -
          mouse-6 v1.1 (Illumina) scl00223664.2_280-S - -
          mouse-6 v1.1 (Illumina) scl0002430.1_79-S - -
          mouse-6 v1.1 (Illumina) scl47785.6_386-S - -
          Illumina Sentrix mouse-6.v1 bead array 106100494 - -
          Illumina Sentrix mouse-6.v1 bead array 6650440 - -
          Illumina Sentrix mouse-6.v1 bead array 5080465 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl00223664.2_280-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0002430.1_79-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6650440 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5080465 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 106100494 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2626606 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2758168 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2626606 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2758168 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0002430.1_79-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline