Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00521722_m1
          See other DICER1 GE Assays ›
          Gene Symbol:
          Dicer1
          Gene Name:
          dicer 1, ribonuclease type III
          Gene Aliases:
          1110006F08Rik, D12Ertd7e, mKIAA0928
          Chromosome Location:
          Chr.12: 104687740 - 104751952 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00521722_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dicer1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_148948.2 NP_683750.2 14 - 15 2506 75
          GenBank mRNA AB081470.1 - 13 - 14 2304 75
          AF430845.1 - 14 - 15 2514 75
          AK129241.1 - 8 - 9 1065 75

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1200002295.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Dicer1

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          meiotic spindle organization
          angiogenesis
          regulation of protein phosphorylation
          hair follicle development
          apoptotic DNA fragmentation
          RNA processing
          cell proliferation
          regulation of Notch signaling pathway
          post-embryonic development
          zygote asymmetric cell division
          regulation of gene expression
          negative regulation of Schwann cell proliferation
          positive regulation of gene expression
          regulation of muscle cell apoptotic process
          positive regulation of Schwann cell differentiation
          myoblast differentiation involved in skeletal muscle regeneration
          rRNA catabolic process
          stem cell population maintenance
          spinal cord motor neuron differentiation
          nerve development
          olfactory bulb interneuron differentiation
          cerebral cortex development
          lung development
          embryonic limb morphogenesis
          production of siRNA involved in RNA interference
          targeting of mRNA for destruction involved in RNA interference
          gene silencing by RNA
          pre-miRNA processing
          hair follicle morphogenesis
          regulation of myelination
          positive regulation of myelination
          peripheral nervous system myelin formation
          conversion of ds siRNA to ss siRNA involved in RNA interference
          siRNA loading onto RISC involved in RNA interference
          embryonic hindlimb morphogenesis
          production of miRNAs involved in gene silencing by miRNA
          multicellular organism growth
          miRNA loading onto RISC involved in gene silencing by miRNA
          conversion of ds siRNA to ss siRNA
          regulation of odontogenesis of dentin-containing tooth
          regulation of viral genome replication
          regulation of cell differentiation
          regulation of neuron differentiation
          positive regulation of transcription from RNA polymerase II promoter
          mRNA stabilization
          spleen development
          digestive tract development
          reproductive structure development
          regulation of oligodendrocyte differentiation
          epidermis morphogenesis
          branching morphogenesis of an epithelial tube
          neuron projection morphogenesis
          anatomical structure development
          regulation of neurogenesis
          cartilage development
          spindle assembly
          regulation of RNA metabolic process
          defense response to virus
          regulation of cell cycle
          cardiac muscle cell development
          inner ear receptor cell development
          negative regulation of glial cell proliferation
          intestinal epithelial cell development
          cardiac neural crest cell development involved in outflow tract morphogenesis
          ganglion development
          regulation of enamel mineralization
          hair follicle cell proliferation
          RNA phosphodiester bond hydrolysis
          RNA phosphodiester bond hydrolysis, endonucleolytic
          regulation of miRNA metabolic process
          positive regulation of miRNA metabolic process
          nucleotide binding
          nucleic acid binding
          RNA binding
          double-stranded RNA binding
          helicase activity
          nuclease activity
          endonuclease activity
          endoribonuclease activity
          ribonuclease III activity
          deoxyribonuclease I activity
          protein binding
          ATP binding
          hydrolase activity
          endoribonuclease activity, producing 5'-phosphomonoesters
          protein domain specific binding
          siRNA binding
          miRNA binding
          metal ion binding
          pre-miRNA binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1200015996.mm.1 NM_148948.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200015996.mm.1 NM_148948.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1200015996.mm.1 TC1200002295.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline