Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00521864_m1
          See other LTC4S GE Assays ›
          Gene Symbol:
          Ltc4s
          Gene Name:
          leukotriene C4 synthase
          Gene Aliases:
          -
          Chromosome Location:
          Chr.11: 50235003 - 50239175 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00521864_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ltc4s
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_008521.1 NP_032547.1 1 - 2 96 60
          GenBank mRNA BC028760.1 - 1 - 2 96 1348372 60
          U27195.1 - 1 - 2 96 60

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100002666.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus Mouse lipid regulated genes Inventoried Mouse_lipid_regulated_genes Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ltc4s

          Panther Classification:

          Molecular Function -

          transferase

          Gene Ontology Categories:

          Function(s) Process(es)

          leukotriene metabolic process
          leukotriene biosynthetic process
          oxidation-reduction process
          glutathione transferase activity
          leukotriene-C4 synthase activity
          glutathione peroxidase activity
          enzyme activator activity
          lipid binding
          lyase activity
          glutathione binding
          protein heterodimerization activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27787506 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27787501 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27787500 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27787498 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27787504 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27787503 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27787502 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27787507 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27787509 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27787510 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2015292 BC028760.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2015292 BC038518.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 136060 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P142308 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2015292 BC028760.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2015292 BC038518.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P354792 NM_008521.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P354792 BC028760.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P354792 BC038518.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P354792 U27195.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27787519 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27787545 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27787522 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27787513 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27787520 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27787511 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27787531 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27787540 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27787534 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27787535 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27787525 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27787514 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27787538 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27787518 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27787501 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27787504 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27787542 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27787507 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27787515 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27787532 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27787506 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27787517 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27787548 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27787516 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27787528 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27787503 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27787498 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27787497 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27787509 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27787499 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27787529 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27787524 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P142308 - -
          Whole Mouse Genome G4122A A_51_P354792 NM_008521.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P354792 BC028760.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P354792 BC038518.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P354792 U27195.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P123736 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P142308 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09061452 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09061449 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09061474 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09061482 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09061444 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09061461 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27787502 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27787498 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27787510 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27787501 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27787504 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27787500 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27787503 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27787509 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27787507 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27787506 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787506 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787441 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787561 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787501 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787429 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787421 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787440 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787498 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787494 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787554 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787562 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787493 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787518 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787529 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787453 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787449 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787522 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787497 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787509 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787434 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787519 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787437 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787504 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787418 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787535 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787534 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787542 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787425 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787525 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787517 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787439 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787548 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787444 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787560 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787420 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787545 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787450 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787451 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787520 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787443 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787433 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787499 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787513 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787447 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787515 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787455 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787422 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787495 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787427 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787511 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787549 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787417 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27787532 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27787518 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27787513 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27787516 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27787548 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27787554 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27787517 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27787511 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27787532 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09061461 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27787507 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09015781 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27787540 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27787546 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09061480 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27787519 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09061447 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27787522 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27787529 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27787552 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27787499 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27787515 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27787495 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27787550 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27787524 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09413713 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27787498 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27787535 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27787534 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27787539 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27787531 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27787557 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27787520 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27787542 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09659328 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27787549 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27787497 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27787538 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10385585 - -
          Affymetrix Mouse Gene 1.0 ST Array 10385584 - -
          Affymetrix Mouse Gene 1.0 ST Array 10385586 - -
          Affymetrix Mouse Gene 1.0 ST Array 10385590 - -
          Affymetrix Mouse Gene 1.0 ST Array 10385588 - -
          Affymetrix Mouse Gene 1.0 ST Array 10385587 - -
          Affymetrix Mouse Gene 1.0 ST Array 10385589 - -
          Affymetrix HT MG-430 PM Array Plate 1419692_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10385583 - -
          Mouse Genome 430 2.0 Array 1419692_a_at - -
          Mouse Genome 430 2.0 Array 1419692_PM_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 17001_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 17001_PM_at - -
          Affymetrix Murine Genome 6500A Array Msa16300_at - -
          Affymetrix Mouse Clariom S Array PSR1100024346.mm.1 NM_008521.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100024346.mm.1 TC1100002666.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100024348.mm.1 NM_008521.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100024348.mm.1 TC1100002666.mm.1 MTA TC
          Mouse Expression Set 430 1419692_a_at - -
          Mouse Expression Set 430 1419692_PM_a_at - -
          Murine Genome U74v2 Set 92401_at - -
          Murine Genome U74v2 Set 92401_PM_at - -
          Murine Genome U74v2 Set 162391_PM_r_at - -
          Murine Genome U74v2 Set 162391_r_at - -
          Mouse Genome 430A 2.0 Array 1419692_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1419692_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1419692_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1419692_PM_a_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2570411 - -
          Illumina mouse-6 v1.0 expression beadchip 2570411 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002570411 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2658687 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2658687 - -
          mouse-6 v1.1 (Illumina) scl40277.4.24_53-S - -
          Illumina Sentrix mouse-6.v1 bead array 2570411 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl40277.4.24_53-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2570411 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2658687 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2658687 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl40277.4.24_53-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline