Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00521920_m1
          See other LECT2 GE Assays ›
          Gene Symbol:
          Lect2
          Gene Name:
          leukocyte cell-derived chemotaxin 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.13: 56528959 - 56548561 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00521920_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lect2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_010702.2 NP_034832.2 2 - 3 314 68
          GenBank mRNA AB009687.1 - 2 - 3 276 68
          AB009688.1 - 2 - 3 185 68
          AF035161.1 - 2 - 3 205 68
          AK131971.1 - 2 - 3 225 68
          BC027753.1 - 2 - 3 279 5049578 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1300002083.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Lect2

          Panther Classification:

          Molecular Function -

          chemokine cytokine intercellular signal molecule

          Gene Ontology Categories:

          Function(s) Process(es)

          chemotaxis
          negative regulation of Wnt signaling pathway
          identical protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2074762 AK131971.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2074762 BC027753.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2074762 AI173435.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2074762 NM_010702.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2074762 AB009687.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 776348 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 770855 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 55066 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 134389 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P166118 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2074762 AK131971.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2074762 BC027753.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2074762 AI173435.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2074762 NM_010702.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2074762 AB009687.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P249909 AK131971.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P249909 BC027753.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P249909 AI173435.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P249909 NM_010702.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P249909 AB009687.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29061961 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29061954 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29061953 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29061958 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29061975 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29061928 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29061962 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29061929 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29061927 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29061969 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29061974 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29061918 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29061934 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29061935 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29061931 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29061959 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29061926 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29061923 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29061951 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29061963 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29061925 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29061972 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29061936 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29061964 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29061952 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29061966 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29061946 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29061939 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29061915 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29061916 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29061920 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29061949 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29061941 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29061944 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29061955 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29061933 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29061913 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29061948 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29061956 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29061937 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07885770 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P166118 - -
          Whole Mouse Genome G4122A A_51_P249909 AK131971.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P249909 BC027753.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P249909 AI173435.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P249909 NM_010702.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P249909 AB009687.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P134914 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P166118 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10863296 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10863287 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10863302 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10863297 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10863312 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10863307 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29061980 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29061949 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29061910 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29061939 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29061953 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29061878 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29061937 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29061925 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29061904 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29061844 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29061967 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29061959 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29061845 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29061956 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29061936 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29061931 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29061927 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29061944 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29061982 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29061894 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29061972 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29061883 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29061923 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29061948 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29061961 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29061896 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29061918 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29061954 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29061929 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29061890 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29061912 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29061986 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29061984 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29061976 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29061966 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29061900 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29061920 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29061908 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29061963 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29061885 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29061958 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29061899 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29061951 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29061974 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29061969 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29061941 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29061933 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29061911 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29061946 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29061978 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29061888 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29061915 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11102555 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29061961 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10863291 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10572783 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29061948 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29061901 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29061944 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29061967 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29061976 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29061936 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11102547 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29061916 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10776581 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29061966 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29061972 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29061888 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29061975 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29061910 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10936771 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29061937 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29061927 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29061963 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10956801 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29061986 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29061900 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29061894 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29061941 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29061908 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29061918 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29061980 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29061911 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29061883 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29061920 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29061897 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29061933 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29061929 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10936770 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29061978 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29061982 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29061974 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11102551 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29061931 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29061904 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29061964 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29061946 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29061885 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29061939 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29061912 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29061934 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29061909 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10621129 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29061923 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10863276 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29061925 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11102554 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29061981 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29061984 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29061969 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29061913 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29061956 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10863314 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29061891 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10409595 - -
          Affymetrix Mouse Gene 1.0 ST Array 10409593 - -
          Affymetrix Mouse Gene 1.0 ST Array 10409596 - -
          Affymetrix Mouse Gene 1.0 ST Array 10409598 - -
          Affymetrix Mouse Gene 1.0 ST Array 10409594 - -
          Affymetrix Mouse Gene 1.0 ST Array 10409597 - -
          Affymetrix HT MG-430 PM Array Plate 1449492_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10409592 - -
          Mouse Genome 430 2.0 Array 1449492_a_at - -
          Mouse Genome 430 2.0 Array 1449492_PM_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 16841_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 16841_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1300013211.mm.1 NM_010702.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013211.mm.1 NM_010702.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013211.mm.1 TC1300002083.mm.1 MTA TC
          Mouse Expression Set 430 1449492_PM_a_at - -
          Mouse Expression Set 430 1449492_a_at - -
          Murine Genome U74v2 Set 102938_at - -
          Murine Genome U74v2 Set 102938_PM_at - -
          Mouse Genome 430A 2.0 Array 1449492_a_at - -
          Mouse Genome 430A 2.0 Array 1449492_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1449492_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1449492_PM_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 660345 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 50279 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000050279 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1223634 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1223634 - -
          mouse-6 v1.1 (Illumina) scl43907.5.1_1-S - -
          Illumina Sentrix mouse-6.v1 bead array 50279 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl43907.5.1_1-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 50279 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1223634 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1223634 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl43907.5.1_1-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline