Search
Search

Limd1| Gene Symbol: Limd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013860.2 | NP_038888.2 | 2 - 3 | 2027 | 91 | |
| GenBank mRNA | AJ132409.1 | - | 2 - 3 | 2016 | 91 | |
| AK004806.1 | - | 2 - 3 | 1331 | 91 | ||
| AK047198.1 | - | 2 - 3 | 2021 | 91 | ||
| AK083970.1 | - | 2 - 3 | 1989 | 91 | ||
| AK180850.1 | - | 1 - 2 | 118 | 91 | ||
| AK193947.1 | - | 1 - 2 | 118 | 91 | ||
| AK194248.1 | - | 1 - 2 | 117 | 91 | ||
| AK197187.1 | - | 1 - 2 | 119 | 91 | ||
| AK207336.1 | - | 1 - 2 | 118 | 91 | ||
| AK210590.1 | - | 1 - 2 | 118 | 91 | ||
| AK217088.1 | - | 1 - 2 | 118 | 91 | ||
| BC056449.1 | - | 2 - 3 | 1902 | 5703026 | 91 | |
Set Membership: |
|