Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00522548_m1
          See other LCN8 GE Assays ›
          Gene Symbol:
          Lcn8
          Gene Name:
          lipocalin 8
          Gene Aliases:
          9230106L18Rik, EP17, Lcn5, mEP17
          Chromosome Location:
          Chr.2: 25653118 - 25656217 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00522548_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lcn8
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_033145.1 NP_149157.1 6 - 7 541 91
          GenBank mRNA AK020323.1 - 3 - 4 149 91
          BC048433.1 - 6 - 7 539 6772515 91

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200000427.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Lcn8

          Panther Classification:

          Molecular Function -

          transfer/carrier protein

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          response to hormone
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2051738 BC048433.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2051738 NM_033145.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 347674 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 212850 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P151702 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2051738 BC048433.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2051738 NM_033145.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P379873 BC048433.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P379873 NM_033145.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21073559 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21073568 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21073610 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21073597 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21073584 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21073586 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21073567 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21073582 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21073563 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21073602 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21073569 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21073577 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21073576 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21073560 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21073591 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21073607 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21073565 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21073603 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21073598 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21073558 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21073605 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21073587 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21073595 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21073570 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21073596 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21073592 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21073614 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21073579 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21073612 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21073590 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21073601 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21073608 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21073600 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21073573 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21073562 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21073566 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21073555 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21073574 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21073561 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21073589 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P151702 - -
          Whole Mouse Genome G4122A A_51_P379873 BC048433.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P379873 NM_033145.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P132111 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P107913 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P151702 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01479875 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01479881 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01479886 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073546 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073591 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073674 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073619 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073641 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073601 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073666 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073543 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073610 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073633 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073603 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073576 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073567 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073643 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073587 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073621 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073573 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073582 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073607 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073647 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073655 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073598 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073677 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073540 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073676 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073635 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073651 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073570 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073675 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073661 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073659 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073637 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073664 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073548 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073648 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073608 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073653 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073658 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073562 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073555 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073624 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073577 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073574 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073649 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073656 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073566 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073612 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073616 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073595 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073669 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073660 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073678 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073600 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073565 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073542 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073640 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073630 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073538 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073545 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073597 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073667 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073559 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073592 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073551 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073605 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073558 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073644 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073645 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073584 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073579 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073665 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073662 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073569 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073589 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073671 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073629 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073625 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073627 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21073605 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01708620 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21073587 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21073608 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21073601 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21073597 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21073555 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21073565 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02241938 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01479897 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21073582 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01708610 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01708623 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01708605 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21073563 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21073600 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21073595 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21073607 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21073589 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21073612 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01479891 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21073562 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21073590 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21073586 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21073569 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21073584 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21073579 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01479875 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21073598 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21073570 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21073573 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21073559 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01479896 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01708612 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21073566 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10470148 - -
          Affymetrix Mouse Gene 1.0 ST Array 10470146 - -
          Affymetrix Mouse Gene 1.0 ST Array 10470145 - -
          Affymetrix Mouse Gene 1.0 ST Array 10470147 - -
          Affymetrix Mouse Gene 1.0 ST Array 10470142 - -
          Affymetrix Mouse Gene 1.0 ST Array 10470144 - -
          Affymetrix Mouse Gene 1.0 ST Array 10470143 - -
          Affymetrix HT MG-430 PM Array Plate 1450609_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10470141 - -
          Mouse Genome 430 2.0 Array 1450609_at - -
          Mouse Genome 430 2.0 Array 1450609_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 78076_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 78076_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0200002607.mm.1 NM_033145.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200002607.mm.1 NM_033145.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200002607.mm.1 TC0200000427.mm.1 MTA TC
          Mouse Expression Set 430 1450609_at - -
          Mouse Expression Set 430 1450609_PM_at - -
          Mouse Genome 430A 2.0 Array 1450609_PM_at - -
          Mouse Genome 430A 2.0 Array 1450609_at - -
          Affymetrix HT Mouse Genome 430A Array 1450609_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1450609_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 584698 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 5720097 - -
          Illumina mouse-6 v1.0 expression beadchip 5720097 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005720097 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2733045 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2733045 - -
          mouse-6 v1.1 (Illumina) scl21170.7.1_39-S - -
          Illumina Sentrix mouse-6.v1 bead array 5720097 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl21170.7.1_39-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5720097 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2733045 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2733045 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline