Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00522934_m1
          See other ANKRD54 GE Assays ›
          Gene Symbol:
          Ankrd54
          Gene Name:
          ankyrin repeat domain 54
          Gene Aliases:
          C730048E16Rik, EST1068184, EST475269, Liar
          Chromosome Location:
          Chr.15: 79053094 - 79062958 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00522934_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ankrd54
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_144849.1 NP_659098.1 2 - 3 437 60
          GenBank mRNA AK133785.1 - 2 - 3 461 60
          AK143625.1 - 2 - 3 464 60
          BC014743.1 - 2 - 3 437 4503319 60
          DQ884878.1 - 2 - 3 465 60

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1500001859.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ankrd54

          Gene Ontology Categories:

          Function(s) Process(es)

          nucleocytoplasmic transport
          positive regulation of erythrocyte differentiation
          regulation of protein kinase activity
          regulation of intracellular signal transduction
          protein binding
          protein kinase regulator activity
          protein complex binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30364126 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30364131 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30364127 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30364129 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30364128 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P307220 AK050440.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P307220 AK050440.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 248869 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 637685 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 334876 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 749821 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 135984 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 722276 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 15331 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 56332 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P114113 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P307220 AK050440.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P307220 AK050440.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30364140 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30364164 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30364137 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30364132 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30364151 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30364159 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30364162 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30364136 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30364145 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30364126 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30364150 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30364141 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30364117 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30364157 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30364134 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30364143 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30364127 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30364123 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30364156 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30364121 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30364124 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30364133 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30364115 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30364147 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30364153 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30364129 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30364166 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30364120 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03078125 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08429259 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P114113 - -
          Whole Mouse Genome G4122A A_51_P307220 AK050440.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P307220 AK050440.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P118424 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P114113 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11946938 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11946935 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11946934 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12311185 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12311187 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30364127 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30364131 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30364128 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30364129 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30364126 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30364008 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30364081 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30364153 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30364102 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30364074 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30364070 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30364137 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30364076 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30364039 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30364142 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30364064 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30364108 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30364007 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30364072 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30364117 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30364014 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30364083 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30364120 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30364126 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30364048 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30364096 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30364077 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30364123 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30364013 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30364162 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30364021 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30364086 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30364156 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30364042 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30364164 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30364058 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30364089 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30364061 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30364159 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30364015 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30364150 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30364073 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30364043 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30364050 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30364129 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30364111 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30364140 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30364101 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30364068 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30364060 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30364033 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30364110 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30364026 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30364127 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30364036 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30364166 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30364133 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30364114 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30364019 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30364031 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30364085 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30364011 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30364132 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30364018 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30364100 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30364099 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30364040 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30364037 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30364066 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30364062 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30364092 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30364136 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30364094 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30364045 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30364105 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30364093 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30364055 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30364134 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12451105 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30364053 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30364066 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30364085 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30364058 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12094410 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30364141 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30364127 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30364083 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11946935 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30364140 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30364120 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30364107 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30364106 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30364064 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30364035 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12422729 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30364105 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30364089 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30364101 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30364142 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30364086 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30364137 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30364126 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30364151 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30364052 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30364102 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30364121 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30364094 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12374303 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30364143 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12195672 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30364060 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30364146 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30364092 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30364114 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30364136 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30364133 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30364093 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30364149 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11984947 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30364145 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11888704 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11888708 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30364062 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30364057 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30364033 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11888706 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30364100 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30364048 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30364134 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11946930 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30364111 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30364055 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30364054 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30364110 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30364123 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10430446 - -
          Affymetrix Mouse Gene 1.0 ST Array 10430439 - -
          Affymetrix Mouse Gene 1.0 ST Array 10430440 - -
          Affymetrix Mouse Gene 1.0 ST Array 10430442 - -
          Affymetrix Mouse Gene 1.0 ST Array 10430441 - -
          Affymetrix Mouse Gene 1.0 ST Array 10430443 - -
          Affymetrix Mouse Gene 1.0 ST Array 10430445 - -
          Affymetrix Mouse Gene 1.0 ST Array 10430444 - -
          Affymetrix HT MG-430 PM Array Plate 1451182_PM_s_at - -
          Affymetrix HT MG-430 PM Array Plate 1438090_PM_x_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10430438 - -
          Mouse Genome 430 2.0 Array 1451182_s_at - -
          Mouse Genome 430 2.0 Array 1438090_x_at - -
          Mouse Genome 430 2.0 Array 1438090_PM_x_at - -
          Mouse Genome 430 2.0 Array 1451182_PM_s_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 223690_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 223690_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1500014508.mm.1 NM_144849.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500014508.mm.1 NM_144849.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500014508.mm.1 TC1500001859.mm.1 MTA TC
          Mouse Expression Set 430 1451182_PM_s_at - -
          Mouse Expression Set 430 1451182_s_at - -
          Mouse Expression Set 430 1438090_PM_x_at - -
          Mouse Expression Set 430 1438090_x_at - -
          Mouse Genome 430A 2.0 Array 1451182_s_at - -
          Mouse Genome 430A 2.0 Array 1438090_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1451182_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1438090_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1438090_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1451182_PM_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1451182_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1438090_PM_x_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 636303 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 3440687 - -
          Illumina mouse-6 v1.0 expression beadchip 3440687 - -
          Illumina mouse-6 v1.0 expression beadchip 100050671 - -
          Illumina mouse-6 v1.0 expression beadchip 1570463 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003440687 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001570463 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2600078 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2722184 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2600078 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2722184 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1248653 - -
          mouse-6 v1.1 (Illumina) scl46973.7_42-S - -
          mouse-6 v1.1 (Illumina) scl0002525.1_37-S - -
          Illumina Sentrix mouse-6.v1 bead array 1570463 - -
          Illumina Sentrix mouse-6.v1 bead array 100050671 - -
          Illumina Sentrix mouse-6.v1 bead array 3440687 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl46973.7_42-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0002525.1_37-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1570463 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 100050671 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3440687 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2722184 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2600078 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2872599 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2579277 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2600078 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2722184 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2872599 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline