Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00523065_m1
          See other SUV420H1 GE Assays ›
          Gene Symbol:
          Suv420h1
          Gene Name:
          suppressor of variegation 4-20 homolog 1 (Drosophila)
          Gene Aliases:
          AA117471, C630029K18Rik, KMT5B, Suv4-20h1
          Chromosome Location:
          Chr.19: 3764155 - 3818303 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00523065_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Suv420h1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001167884.1 NP_001161356.1 7 - 8 1233 68
          NM_001167885.1 NP_001161357.1 4 - 5 931 68
          NM_001167886.1 NP_001161358.1 3 - 4 871 68
          NM_001167887.1 NP_001161359.1 3 - 4 871 68
          NM_001167888.1 NP_001161360.1 4 - 5 931 68
          NM_001167889.1 NP_001161361.1 3 - 4 871 68
          NM_144871.4 NP_659120.3 4 - 5 931 68
          GenBank mRNA AK054093.1 - 2 - 3 467 68
          AK082660.1 - 2 - 3 551 68
          AK083227.1 - 4 - 5 932 68
          AK087267.1 - 7 - 8 1135 68
          AK139255.1 - 2 - 3 436 68
          AK152179.1 - 3 - 4 475 68
          AY555192.1 - 2 - 3 561 68
          BC011214.1 - 3 - 4 497 68
          BC075709.1 - 2 - 3 3661032 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1900000017.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Suv420h1

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          muscle organ development
          chromatin modification
          histone methylation
          methylation
          histone H4-K20 methylation
          histone H4-K20 trimethylation
          histone lysine methylation
          methyltransferase activity
          transferase activity
          histone-lysine N-methyltransferase activity
          histone methyltransferase activity (H4-K20 specific)

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1900000117.mm.1 NM_001167884.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000117.mm.1 NM_001167884.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000117.mm.1 NM_001167885.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000117.mm.1 NM_001167885.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000117.mm.1 NM_001167888.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000117.mm.1 NM_001167888.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000117.mm.1 NM_144871.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000117.mm.1 NM_144871.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000117.mm.1 TC1900000017.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900000115.mm.1 NM_001167884.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000115.mm.1 NM_001167884.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000115.mm.1 TC1900000017.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900000119.mm.1 NM_001167884.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000119.mm.1 NM_001167884.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000119.mm.1 TC1900000017.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900000124.mm.1 NM_001167884.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000124.mm.1 NM_001167884.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000124.mm.1 NM_001167885.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000124.mm.1 NM_001167885.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000124.mm.1 NM_001167886.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000124.mm.1 NM_001167886.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000124.mm.1 NM_001167887.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000124.mm.1 NM_001167887.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000124.mm.1 NM_001167888.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000124.mm.1 NM_001167888.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000124.mm.1 NM_001167889.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000124.mm.1 NM_001167889.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000124.mm.1 NM_144871.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000124.mm.1 NM_144871.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000124.mm.1 TC1900000017.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline