Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00523393_m1
          See other TMEM150A GE Assays ›
          Gene Symbol:
          Tmem150a
          Gene Name:
          transmembrane protein 150A
          Gene Aliases:
          BC014685, Tmem150
          Chromosome Location:
          Chr.6: 72355483 - 72359762 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00523393_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tmem150a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_144916.3 NP_659165.1 3 - 4 290 74
          GenBank mRNA BC014685.1 - 2 - 3 206 4241375 74
          BC028825.1 - 3 - 4 292 5097962 74

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600000849.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tmem150a

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          regulation of autophagy
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24541278 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24541281 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24541279 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24541280 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24541275 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P164939 NM_144916.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P164939 BC014685.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P164939 BC028825.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P164939 NM_144916.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P164939 BC014685.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P164939 BC028825.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 539701 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 479473 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P164939 NM_144916.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P164939 BC014685.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P164939 BC028825.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P164939 NM_144916.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P164939 BC014685.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P164939 BC028825.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24541283 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24541279 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24541281 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24541290 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24541275 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24541299 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24541285 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24541284 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24541277 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24541286 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24541301 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24541288 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24541294 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24541297 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24541292 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24541278 - -
          Whole Mouse Genome G4122A A_51_P164939 NM_144916.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P164939 BC014685.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P164939 BC028825.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P164939 NM_144916.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P164939 BC014685.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P164939 BC028825.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P131718 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05619247 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05619232 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05619238 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24541281 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24541275 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24541278 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24541279 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24541280 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541288 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541283 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541328 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541211 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541335 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541297 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541232 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541238 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541343 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541216 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541324 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541294 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541243 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541245 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541222 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541208 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541327 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541267 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541207 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541268 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541252 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541317 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541248 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541234 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541277 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541290 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541221 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541274 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541240 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541299 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541229 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541249 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541311 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541284 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541244 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541258 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541257 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541262 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541264 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541235 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541218 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541350 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541352 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541231 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541220 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541330 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541332 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541266 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541255 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541326 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541272 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541336 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541354 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541292 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541214 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541345 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541260 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541320 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541346 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541250 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541210 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541322 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541217 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541279 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541305 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541281 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541278 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541275 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541301 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541319 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541225 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541329 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541270 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541302 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541323 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541236 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541228 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541316 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541286 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541224 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541307 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24541206 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24541259 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24541275 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24541262 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24541281 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05580939 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24541245 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24541244 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24541272 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24541279 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05716367 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05313603 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24541243 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24541292 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24541302 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24541255 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24541257 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24541238 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24541240 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24541258 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24541265 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24541290 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05716370 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24541297 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24541278 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24541305 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24541306 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05716379 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24541288 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05619238 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24541294 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05580947 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24541299 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24541286 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24541283 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05075283 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24541252 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24541311 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05716375 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24541260 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24541307 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24541308 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10539118 - -
          Affymetrix Mouse Gene 1.0 ST Array 10539115 - -
          Affymetrix Mouse Gene 1.0 ST Array 10539117 - -
          Affymetrix Mouse Gene 1.0 ST Array 10539114 - -
          Affymetrix Mouse Gene 1.0 ST Array 10539112 - -
          Affymetrix Mouse Gene 1.0 ST Array 10539116 - -
          Affymetrix Mouse Gene 1.0 ST Array 10539113 - -
          Affymetrix HT MG-430 PM Array Plate 1424726_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10539111 - -
          Mouse Genome 430 2.0 Array 1424726_PM_at - -
          Mouse Genome 430 2.0 Array 1424726_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 232086_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 232086_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0600006487.mm.1 TC0600000849.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600006487.mm.1 NM_144916.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600006487.mm.1 NM_144916.3 Refseq
          Mouse Expression Set 430 1424726_at - -
          Mouse Expression Set 430 1424726_PM_at - -
          Murine Genome U74v2 Set 103078_PM_at - -
          Murine Genome U74v2 Set 103078_at - -
          Mouse Genome 430A 2.0 Array 1424726_PM_at - -
          Mouse Genome 430A 2.0 Array 1424726_at - -
          Affymetrix HT Mouse Genome 430A Array 1424726_at - -
          Affymetrix HT Mouse Genome 430A Array 1424726_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 636279 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 2120605 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002120605 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2653396 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2653396 - -
          mouse-6 v1.1 (Illumina) scl29884.7.1_23-S - -
          Illumina Sentrix mouse-6.v1 bead array 2120605 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl29884.7.1_23-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2120605 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2653395 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2861123 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2653395 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2861123 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline