Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00523472_m1
          See other TBC1D17 GE Assays ›
          Gene Symbol:
          Tbc1d17
          Gene Name:
          TBC1 domain family, member 17
          Gene Aliases:
          BC017607
          Chromosome Location:
          Chr.7: 44840776 - 44849079 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00523472_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tbc1d17
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001042655.1 NP_001036120.1 7 - 8 1010 112
          GenBank mRNA AK039566.1 - 7 - 8 825 112
          AK046883.1 - 7 - 8 844 112
          AK075571.1 - 1 - 2 114 112
          AK081318.1 - 7 - 8 823 112
          AK083153.1 - 7 - 8 828 112
          AK136705.1 - 7 - 8 835 112
          AK149917.1 - 7 - 8 822 112
          AK153732.1 - 7 - 8 822 112
          AK170821.1 - 7 - 8 822 112
          BC017607.1 - 7 - 8 1010 4507589 112
          BC035449.1 - 7 - 8 809 112

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700002975.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the AK046883.1 transcripts.

          Set Membership:

          Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          GTPase-activating protein

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          autophagy
          protein transport
          retrograde transport, endosome to Golgi
          molecular_function
          GTPase activator activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25089659 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25089655 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25089658 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25089654 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25089657 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25089660 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25089653 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25089652 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P655084 BC017607.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P655084 BC035449.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P655084 AK153732.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P655084 AK170821.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P655084 AK083153.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P655084 BC017607.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P655084 BC035449.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P655084 AK153732.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P655084 AK170821.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P655084 AK083153.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 598974 AK046303.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 598974 AK155745.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 598974 AK170937.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 598974 BC132379.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 598974 NM_001111058.1 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 598974 NM_021293.3 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 598974 S71403.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 807853 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 123294 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 222694 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 3891 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 286048 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P126088 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P140301 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P655084 BC017607.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P655084 BC035449.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P655084 AK153732.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P655084 AK170821.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P655084 AK083153.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P655084 BC017607.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P655084 BC035449.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P655084 AK153732.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P655084 AK170821.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P655084 AK083153.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P655084 BC017607.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P655084 BC035449.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P655084 AK153732.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P655084 AK170821.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P655084 AK083153.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P655084 BC017607.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P655084 BC035449.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P655084 AK153732.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P655084 AK170821.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P655084 AK083153.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P283954 AK087754.1 mouse_transcriptome
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06284634 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06284621 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06284624 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06284629 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06284638 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25089659 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25089655 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25089654 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25089658 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25089660 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25089657 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25089653 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25089652 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089625 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089521 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089637 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089633 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089583 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089551 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089540 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089587 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089559 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089645 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089612 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089532 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089634 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089606 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089647 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089572 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089638 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089600 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089580 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089574 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089556 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089533 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089654 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089576 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089525 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089566 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089655 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089650 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089545 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089640 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089578 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089577 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089520 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089581 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089631 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089658 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089534 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089630 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089660 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089571 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089549 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089614 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089562 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089643 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089548 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089642 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089553 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089602 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089568 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089599 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089569 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089589 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089619 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089649 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089627 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089544 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089527 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089611 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089644 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089564 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089558 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089541 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089609 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089573 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089616 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089523 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089579 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089575 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089624 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089595 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089585 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089632 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089591 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089605 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089543 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089560 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089593 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25089622 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25089633 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25089625 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25089649 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25089609 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25089647 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25089631 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25089644 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25089632 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06141736 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25089612 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25089658 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25089650 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25089595 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25089622 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25089614 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25089611 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25089655 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25089602 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25089605 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06101725 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06284643 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25089606 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25089624 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25089627 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25089628 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25089616 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25089640 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25089636 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25089645 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25089638 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25089629 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25089654 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06559014 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25089634 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25089642 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25089619 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10562922 - -
          Affymetrix Mouse Gene 1.0 ST Array 10562914 - -
          Affymetrix Mouse Gene 1.0 ST Array 10562923 - -
          Affymetrix Mouse Gene 1.0 ST Array 10562919 - -
          Affymetrix Mouse Gene 1.0 ST Array 10562917 - -
          Affymetrix Mouse Gene 1.0 ST Array 10562915 - -
          Affymetrix Mouse Gene 1.0 ST Array 10562918 - -
          Affymetrix Mouse Gene 1.0 ST Array 10562926 - -
          Affymetrix Mouse Gene 1.0 ST Array 10562920 - -
          Affymetrix Mouse Gene 1.0 ST Array 10562913 - -
          Affymetrix Mouse Gene 1.0 ST Array 10562921 - -
          Affymetrix Mouse Gene 1.0 ST Array 10562912 - -
          Affymetrix Mouse Gene 1.0 ST Array 10562925 - -
          Affymetrix Mouse Gene 1.0 ST Array 10562924 - -
          Affymetrix Mouse Gene 1.0 ST Array 10562916 - -
          Affymetrix HT MG-430 PM Array Plate 1423659_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10562911 - -
          Mouse Genome 430 2.0 Array 1423659_PM_a_at - -
          Mouse Genome 430 2.0 Array 1423659_a_at - -
          Affymetrix Mouse Clariom S Array PSR0700026533.mm.1 NM_001042655.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700026533.mm.1 NM_001042655.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700026533.mm.1 TC0700002975.mm.1 MTA TC
          Mouse Expression Set 430 1423659_PM_a_at - -
          Mouse Expression Set 430 1423659_a_at - -
          Mouse Genome 430A 2.0 Array 1423659_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1423659_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1423659_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1423659_PM_a_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 105900465 - -
          MouseWG-6 v2 Expression BeadChip 2320050 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2611993 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2611993 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2611993 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline