| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P25089652 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P25089657 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P25089654 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P25089655 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P25089659 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P25089660 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P25089653 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P25089658 |
- |
- |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P655084 |
BC017607.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P655084 |
BC035449.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P655084 |
AK153732.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P655084 |
AK170821.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P655084 |
AK083153.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P655084 |
BC017607.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P655084 |
BC035449.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P655084 |
AK153732.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P655084 |
AK170821.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P655084 |
AK083153.1 |
mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
807853 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
3891 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
123294 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
286048 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
598974 |
AK046303.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
598974 |
AK155745.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
598974 |
AK170937.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
598974 |
BC132379.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
598974 |
NM_001111058.1 |
refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
598974 |
NM_021293.3 |
refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
598974 |
S71403.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
222694 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P140301 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P126088 |
- |
- |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P655084 |
BC017607.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P655084 |
BC035449.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P655084 |
AK153732.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P655084 |
AK170821.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P655084 |
AK083153.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P655084 |
BC017607.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P655084 |
BC035449.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P655084 |
AK153732.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P655084 |
AK170821.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P655084 |
AK083153.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P283954 |
AK087754.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P655084 |
BC017607.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P655084 |
BC035449.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P655084 |
AK153732.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P655084 |
AK170821.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P655084 |
AK083153.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P655084 |
BC017607.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P655084 |
BC035449.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P655084 |
AK153732.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P655084 |
AK170821.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P655084 |
AK083153.1 |
mouse_transcriptome |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P06284638 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P06284624 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P06284634 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P06284629 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P06284621 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P25089654 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P25089655 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P25089658 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P25089653 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P25089659 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P25089657 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P25089660 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P25089652 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25089541 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25089593 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25089630 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25089534 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25089599 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25089571 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25089551 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25089559 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25089612 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25089638 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25089548 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25089602 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25089655 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25089556 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25089633 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25089558 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25089591 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25089634 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25089527 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25089523 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25089578 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25089581 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25089650 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25089568 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25089521 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25089549 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25089564 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25089575 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25089577 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25089614 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25089569 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25089619 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25089532 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25089644 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25089562 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25089643 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25089543 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25089533 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25089631 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25089566 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25089572 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25089660 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25089540 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25089576 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25089544 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25089645 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25089560 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25089654 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25089573 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25089587 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25089606 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25089600 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25089627 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25089605 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25089589 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25089640 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25089642 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25089574 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25089579 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25089585 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25089616 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25089611 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25089624 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25089609 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25089649 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25089658 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25089580 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25089622 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25089647 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25089553 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25089545 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25089637 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25089595 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25089520 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25089625 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25089583 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25089525 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25089632 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25089638 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25089606 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25089611 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25089650 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25089609 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25089632 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25089633 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25089628 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25089642 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06101725 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25089602 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25089645 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25089647 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25089616 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25089614 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25089605 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25089636 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25089622 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25089640 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25089649 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25089624 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06141736 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25089619 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25089595 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25089634 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06559014 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25089627 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25089654 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25089644 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25089625 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06284643 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25089631 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25089655 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25089612 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25089629 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25089658 |
- |
- |