Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00523523_m1
          See other MBD3L2 GE Assays ›
          Gene Symbol:
          Mbd3l2
          Gene Name:
          methyl-CpG binding domain protein 3-like 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.9: 18430615 - 18446316 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00523523_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mbd3l2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_144934.3 NP_659183.3 2 - 3 260 103
          GenBank mRNA AF503920.1 - 1 - 2 94 103
          AK135772.1 - 2 - 3 251 103
          BC064744.1 - 2 - 3 221 30051497 103

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900000135.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mbd3l2

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2172500 BC064744.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2172500 NM_144934.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 729520 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 273565 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 696339 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 288510 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 555692 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 295755 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 309964 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 148213 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 152570 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P159721 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2172500 BC064744.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2172500 NM_144934.3 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26336846 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26336851 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26336855 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26336852 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26336849 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26336845 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26336857 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26336842 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26336856 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26336836 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26336858 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26336847 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26336853 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P159721 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06713470 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06713478 - -
          Whole Mouse Genome G4122A A_51_P498042 BC064744.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P498042 NM_144934.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P498042 AF503920.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P134465 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P136195 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P159721 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08162411 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08144195 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26336917 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26336908 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26336922 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26336924 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26336826 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26336861 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26336916 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26336946 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26336844 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26336962 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26336894 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26336889 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26336909 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26336948 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26336933 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26336947 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26336851 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26336847 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26336899 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26336864 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26336921 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26336824 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26336878 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26336853 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26336891 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26336964 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26336869 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26336874 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26336886 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26336897 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26336906 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26336956 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26336935 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26336896 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26336863 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26336959 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26336846 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26336871 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26336905 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26336952 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26336914 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26336902 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26336855 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26336858 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26336852 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26336882 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26336883 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26336879 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26336960 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26336938 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26336934 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26336857 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26336857 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26336842 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26336844 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07815591 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26336873 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26336836 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08102747 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26336839 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26336874 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26336864 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08144195 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07815592 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26336861 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26336852 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26336878 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26336855 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26336866 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26336851 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26336863 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08102746 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26336862 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26336869 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26336847 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26336846 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26336858 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26336849 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08102750 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07698133 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26336853 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10583366 - -
          Affymetrix Mouse Gene 1.0 ST Array 10583369 - -
          Affymetrix Mouse Gene 1.0 ST Array 10583368 - -
          Affymetrix Mouse Gene 1.0 ST Array 10583370 - -
          Affymetrix Mouse Gene 1.0 ST Array 10583367 - -
          Affymetrix HT MG-430 PM Array Plate 1459988_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1452644_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10583365 - -
          Mouse Genome 430 2.0 Array 1452644_PM_at - -
          Mouse Genome 430 2.0 Array 1452644_at - -
          Mouse Genome 430 2.0 Array 1459988_PM_at - -
          Mouse Genome 430 2.0 Array 1459988_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 234988_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 234988_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0900000947.mm.1 NM_144934.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900000947.mm.1 NM_144934.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0900000947.mm.1 TC0900000135.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900000951.mm.1 NM_144934.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900000951.mm.1 NM_144934.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0900000951.mm.1 TC0900000135.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900000948.mm.1 NM_144934.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900000948.mm.1 NM_144934.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0900000948.mm.1 TC0900000135.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900000950.mm.1 NM_144934.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900000950.mm.1 NM_144934.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0900000950.mm.1 TC0900000135.mm.1 MTA TC
          Mouse Expression Set 430 1452644_at - -
          Mouse Expression Set 430 1452644_PM_at - -
          Mouse Expression Set 430 1459988_PM_at - -
          Mouse Expression Set 430 1459988_at - -
          Murine Genome U74v2 Set 133050_at - -
          Murine Genome U74v2 Set 133050_PM_at - -
          Mouse Genome 430A 2.0 Array 1452644_PM_at - -
          Mouse Genome 430A 2.0 Array 1452644_at - -
          Affymetrix HT Mouse Genome 430A Array 1452644_at - -
          Affymetrix HT Mouse Genome 430A Array 1452644_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1459988_at - -
          Affymetrix HT Mouse Genome 430B Array 1459988_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 517537 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 430446 - -
          Illumina mouse-6 v1.0 expression beadchip 3120309 - -
          Illumina mouse-6 v1.0 expression beadchip 430446 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000430446 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003120309 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2609129 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2751997 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2751997 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2609129 - -
          mouse-6 v1.1 (Illumina) scl0234988.3_288-S - -
          mouse-6 v1.1 (Illumina) scl37258.3.1_3-S - -
          Illumina Sentrix mouse-6.v1 bead array 430446 - -
          Illumina Sentrix mouse-6.v1 bead array 3120309 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl37258.3.1_3-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0234988.3_288-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3120309 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 430446 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2751997 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2609129 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2609129 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2751997 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0234988.3_288-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline