Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00523545_m1
          See other CD207 GE Assays ›
          Gene Symbol:
          Cd207
          Gene Name:
          CD207 antigen
          Gene Aliases:
          -
          Chromosome Location:
          Chr.6: 83671207 - 83677857 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00523545_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cd207
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_144943.3 NP_659192.2 4 - 5 784 69
          GenBank mRNA AJ302711.1 - 4 - 5 784 69
          AK142731.1 - 4 - 5 726 69
          AY026050.1 - 4 - 5 726 69
          AY078415.1 - 4 - 5 726 69

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600002579.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cd207

          Panther Classification:

          Molecular Function -

          membrane traffic protein

          Gene Ontology Categories:

          Function(s) Process(es)

          defense response to virus
          carbohydrate binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P110381 AJ302711.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P110381 AY026050.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P110381 AJ302711.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P110381 AY026050.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 49657 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 968266 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 373138 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 327430 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 608275 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 109101 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P107880 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P110381 AJ302711.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P110381 AY026050.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P110381 AJ302711.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P110381 AY026050.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P110381 AJ302711.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P110381 AY026050.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P110381 AJ302711.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P110381 AY026050.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24594836 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24594831 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24594866 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24594852 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24594842 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24594863 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24594840 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24594867 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24594864 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24594839 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24594841 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24594829 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24594846 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24594833 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24594845 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24594869 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24594871 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24594827 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24594849 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24594862 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24594834 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24594860 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24594857 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24594844 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24594858 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24594855 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24594848 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24594850 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P107880 - -
          Whole Mouse Genome G4122A A_51_P110381 AJ302711.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P110381 AY026050.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P110381 AJ302711.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P110381 AY026050.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P105670 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P107880 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05427216 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05427220 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05427214 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05427208 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05427221 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05098771 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24594752 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24594754 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24594845 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24594781 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24594883 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24594850 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24594819 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24594777 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24594774 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24594869 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24594764 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24594879 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24594771 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24594857 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24594808 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24594792 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24594798 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24594862 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24594833 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24594794 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24594758 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24594846 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24594895 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24594863 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24594805 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24594755 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24594831 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24594827 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24594775 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24594785 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24594840 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24594875 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24594751 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24594823 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24594759 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24594763 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24594821 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24594795 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24594886 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24594829 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24594849 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24594773 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24594871 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24594856 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24594864 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24594789 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24594810 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24594897 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24594839 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24594750 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24594790 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24594812 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24594765 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24594836 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24594800 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24594809 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24594756 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24594782 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24594807 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24594877 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24594855 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24594881 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24594748 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24594802 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24594888 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24594860 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24594768 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24594778 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24594894 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24594814 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24594770 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24594885 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24594761 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24594867 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24594873 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24594842 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24594893 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24594816 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05063081 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24594795 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05411094 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24594805 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24594812 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24594860 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24594840 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24594836 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24594829 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05651404 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24594885 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24594823 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24594863 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24594862 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24594846 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24594849 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05383271 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05555704 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24594798 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05063083 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24594801 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24594883 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24594833 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24594866 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05185213 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05624613 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24594837 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05624610 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05651402 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24594845 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05353700 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24594864 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24594855 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24594810 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24594800 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05719180 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24594796 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05353701 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05063084 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24594819 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05796808 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05796809 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24594886 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24594892 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24594806 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24594816 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24594875 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05427214 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05314483 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05413912 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24594793 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24594879 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24594881 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24594802 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05796814 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24594852 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24594871 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24594839 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05427216 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24594888 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05485134 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10545747 - -
          Affymetrix Mouse Gene 1.0 ST Array 10545749 - -
          Affymetrix Mouse Gene 1.0 ST Array 10545750 - -
          Affymetrix Mouse Gene 1.0 ST Array 10545745 - -
          Affymetrix Mouse Gene 1.0 ST Array 10545748 - -
          Affymetrix Mouse Gene 1.0 ST Array 10545744 - -
          Affymetrix Mouse Gene 1.0 ST Array 10545746 - -
          Affymetrix Mouse Gene 1.0 ST Array 10545743 - -
          Affymetrix HT MG-430 PM Array Plate 1425243_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10545742 - -
          Mouse Genome 430 2.0 Array 1425243_PM_at - -
          Mouse Genome 430 2.0 Array 1425243_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 246278_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 246278_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0600020031.mm.1 TC0600002579.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600020031.mm.1 NM_144943.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600020031.mm.1 NM_144943.3 Refseq
          Mouse Expression Set 430 1425243_at - -
          Mouse Expression Set 430 1425243_PM_at - -
          Mouse Genome 430A 2.0 Array 1425243_PM_at - -
          Mouse Genome 430A 2.0 Array 1425243_at - -
          Affymetrix HT Mouse Genome 430A Array 1425243_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1425243_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 834531 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 1990139 - -
          Illumina mouse-6 v1.0 expression beadchip 1990139 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001990139 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2591543 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2591543 - -
          mouse-6 v1.1 (Illumina) scl28790.6.1_57-S - -
          Illumina Sentrix mouse-6.v1 bead array 1990139 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl28790.6.1_57-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1990139 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2785648 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2591543 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2591543 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2785648 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline