Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00523556_s1
          See other HIST1H3G GE Assays ›
          Gene Symbol:
          Hist1h3g
          Gene Name:
          histone cluster 1, H3g
          Gene Aliases:
          H3.1-221, M32460
          Chromosome Location:
          Chr.13: 23535418 - 23535909 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00523556_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Hist1h3g
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_145073.2 NP_659539.1 1 433 93
          GenBank mRNA BC125355.1 - 1 429 40129870 93
          BC125357.1 - 1 429 40129872 93

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1300000246.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Hist1h3g

          Panther Classification:

          Molecular Function -

          histone

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of defense response to virus by host
          nucleosome assembly
          DNA replication-dependent nucleosome assembly
          protein heterotetramerization
          regulation of gene silencing
          histone binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28882028 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28882031 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28882027 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2068260 NM_145073.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2068260 BC125355.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2068260 BC125357.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2068260 NM_145073.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2068260 BC125355.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2068260 BC125357.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28882025 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28882021 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28882027 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28882031 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28882028 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28882019 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28882022 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P100925 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10622618 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10743304 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10622611 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10743303 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28882027 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28882031 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28882028 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882005 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28881986 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28881966 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28881968 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28881991 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882021 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28881975 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28881999 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28881982 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28881989 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28881993 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28881957 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28881964 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882006 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882000 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28881979 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882003 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28881994 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28881969 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882059 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882068 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882042 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882002 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28881996 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882025 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882013 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28881981 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28881992 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882038 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28881958 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882028 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882033 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882062 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882034 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882037 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28881967 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882041 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882031 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28881972 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28881987 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882054 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882027 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882057 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882019 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882074 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28881980 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28881960 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882011 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28881988 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882075 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28882027 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28882021 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28882025 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10622616 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28882019 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28882042 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28882039 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28882033 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10585503 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28882043 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28882037 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28882050 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10603854 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28882038 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28882022 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28882034 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28882028 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10404050 - -
          Affymetrix Mouse Gene 1.0 ST Array 10408084 - -
          Affymetrix Mouse Gene 1.0 ST Array 10403942 - -
          Affymetrix Mouse Gene 1.0 ST Array 10404029 - -
          Affymetrix Mouse Gene 1.0 ST Array 10408248 - -
          Affymetrix Mouse Gene 1.1 ST Array 10408246 - -
          Affymetrix Mouse Gene 1.1 ST Array 10404028 - -
          Affymetrix Mouse Gene 1.1 ST Array 10403941 - -
          Affymetrix Mouse Gene 1.1 ST Array 10408083 - -
          Affymetrix Mouse Clariom S Array PSR1300001400.mm.1 NM_145073.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300001400.mm.1 NM_145073.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1300001400.mm.1 TC1300000246.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300001401.mm.1 TC1300000246.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300001399.mm.1 NM_145073.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300001399.mm.1 NM_145073.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1300001399.mm.1 TC1300000246.mm.1 MTA TC
          Mouse Expression Set 430 1427864_PM_at - -
          Mouse Expression Set 430 1460314_s_at - -
          Mouse Expression Set 430 1460314_PM_s_at - -
          Mouse Expression Set 430 1427864_at - -
          Murine Genome U74v2 Set 93024_PM_at - -
          Murine Genome U74v2 Set 101784_f_at - -
          Murine Genome U74v2 Set 101784_PM_f_at - -
          Murine Genome U74v2 Set 93023_f_at - -
          Murine Genome U74v2 Set 93024_at - -
          Murine Genome U74v2 Set 96416_PM_f_at - -
          Murine Genome U74v2 Set 101783_PM_i_at - -
          Murine Genome U74v2 Set 96416_f_at - -
          Murine Genome U74v2 Set 94756_at - -
          Murine Genome U74v2 Set 94756_PM_at - -
          Murine Genome U74v2 Set 101783_i_at - -
          Murine Genome U74v2 Set 93023_PM_f_at - -
          Mouse Genome 430A 2.0 Array 1460314_s_at - -
          Mouse Genome 430A 2.0 Array 1460314_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1427864_PM_at - -
          Mouse Genome 430A 2.0 Array 1427864_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 5690524 - -
          Illumina mouse-6 v1.0 expression beadchip 5690524 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005690524 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2777961 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2777961 - -
          mouse-6 v1.1 (Illumina) scl097908.1_77-S - -
          Illumina Sentrix mouse-6.v1 bead array 5690524 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl097908.1_77-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5690524 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3002290 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2777961 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3002290 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline