Search
Search
| Gene Symbol: Gbp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083312.2 | NP_001076781.1 | 5 - 6 | 741 | 71 | |
| NM_145545.4 | NP_663520.2 | 5 - 6 | 759 | 71 | ||
| GenBank mRNA | AK036666.1 | - | 5 - 6 | 755 | 71 | |
| AK087751.1 | - | 5 - 6 | 735 | 71 | ||
| AK131176.1 | - | 4 - 5 | 642 | 71 | ||
| BC010229.1 | - | 5 - 6 | 706 | 3501441 | 71 | |
Set Membership: |
|