Search
Search

Ccdc136| Gene Symbol: Ccdc136 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001201378.1 | NP_001188307.1 | 6 - 7 | 1032 | 69 | |
| NM_145574.3 | NP_663549.3 | 6 - 7 | 1032 | 69 | ||
| GenBank mRNA | BC006583.1 | - | 4 - 5 | 620 | 69 | |
| CX222432.1 | - | 3 - 4 | 592 | 69 | ||
| CX564275.1 | - | 3 - 4 | 537 | 69 | ||
| EU029664.1 | - | 6 - 7 | 1032 | 69 | ||
| EU029665.1 | - | 6 - 7 | 1032 | 69 | ||
Set Membership: |
|