| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Gene 1.0 ST Array |
10559879 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10559924 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10549936 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10549937 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10559927 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10559836 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10559920 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10559832 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10559834 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10559921 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10559833 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10559835 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10549933 |
- |
- |
| Affymetrix HT MG-430 PM Array Plate |
1437567_PM_at |
- |
- |
| Affymetrix HT MG-430 PM Array Plate |
1460381_PM_at |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10559919 |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10549932 |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10559878 |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10559831 |
- |
- |
| Mouse Genome 430 2.0 Array |
1460381_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1460381_PM_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1437567_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1437567_PM_at |
- |
- |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array |
232855_PM_at |
- |
- |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array |
232855_at |
- |
- |
| Affymetrix Mouse Clariom S Array |
PSR0700020177.mm.1 |
TC0700002223.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700020177.mm.1 |
NM_145577.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700020177.mm.1 |
NM_145577.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700020176.mm.1 |
TC0700002223.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700020176.mm.1 |
NM_145577.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700020176.mm.1 |
NM_145577.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700020179.mm.1 |
TC0700002223.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700020179.mm.1 |
NM_145577.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700020179.mm.1 |
NM_145577.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700020178.mm.1 |
TC0700002223.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700020178.mm.1 |
NM_145577.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700020178.mm.1 |
NM_145577.2 |
Refseq |
| Mouse Expression Set 430 |
1460381_at |
- |
- |
| Mouse Expression Set 430 |
1460381_PM_at |
- |
- |
| Mouse Expression Set 430 |
1437567_PM_at |
- |
- |
| Mouse Expression Set 430 |
1437567_at |
- |
- |
| Murine Genome U74v2 Set |
135632_PM_f_at |
- |
- |
| Murine Genome U74v2 Set |
135632_f_at |
- |
- |
| Mouse Genome 430A 2.0 Array |
1460381_PM_at |
- |
- |
| Mouse Genome 430A 2.0 Array |
1460381_at |
- |
- |
| Affymetrix HT Mouse Genome 430A Array |
1460381_PM_at |
- |
- |
| Affymetrix HT Mouse Genome 430A Array |
1460381_at |
- |
- |
| Affymetrix HT Mouse Genome 430B Array |
1437567_at |
- |
- |
| Affymetrix HT Mouse Genome 430B Array |
1437567_PM_at |
- |
- |