Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00524088_m1
          See other RPP40 GE Assays ›
          Gene Symbol:
          Rpp40
          Gene Name:
          ribonuclease P 40 subunit
          Gene Aliases:
          D8Bwg1265e, Rnasep1
          Chromosome Location:
          Chr.13: 35895104 - 35911565 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00524088_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rpp40
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_145938.4 NP_666050.3 7 - 8 912 107
          GenBank mRNA AK048810.1 - 912 107
          BC024607.1 - 7 - 8 857 4236826 107
          BC037666.1 - 5 - 6 3055 107

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1300001804.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rpp40

          Gene Ontology Categories:

          Function(s) Process(es)

          tRNA 5'-leader removal
          tRNA processing
          RNA phosphodiester bond hydrolysis, endonucleolytic
          ribonuclease P activity
          hydrolase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28945956 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28945954 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P366047 BC024607.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P366047 NM_145938.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P366047 AK048810.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P366047 BC024607.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P366047 NM_145938.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P366047 AK048810.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 967531 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 656204 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 337513 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 241300 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 873315 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 416908 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 827605 AK051764.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 844399 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 883072 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 32508 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P129662 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P175890 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P366047 BC024607.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P366047 NM_145938.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P366047 AK048810.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P366047 BC024607.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P366047 NM_145938.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P366047 AK048810.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P319392 AK048810.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28945953 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28945972 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28945943 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28945965 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28945945 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28945938 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28945955 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28945969 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28945957 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28945964 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28945940 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28945950 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28945962 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28945970 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28945947 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28945952 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28945939 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28945948 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28945968 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28945959 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P129662 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07836779 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07836748 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P175890 - -
          Whole Mouse Genome G4122A A_51_P319392 AK048810.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P366047 BC024607.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P366047 NM_145938.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P366047 AK048810.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P366047 BC024607.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P366047 NM_145938.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P366047 AK048810.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P130169 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P109674 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P175890 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P129662 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10456143 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10873740 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10873738 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10593919 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10593926 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28945956 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28945954 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28945877 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28945892 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28945898 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28945986 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28945932 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28945886 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28945970 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28945945 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28945955 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28945950 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28945959 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28945905 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28945953 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28945845 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28945904 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28945980 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28945888 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28945968 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28945947 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28945843 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28945863 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28945858 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28945879 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28945890 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28945923 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28945943 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28945984 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28945850 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28945921 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28945938 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28945881 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28945906 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28945854 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28945885 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28945847 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28945853 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28945977 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28945910 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28945940 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28945857 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28945887 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28945916 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28945846 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28945912 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28945939 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28945837 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28945931 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28945873 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28945894 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28945907 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28945842 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28945971 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28945915 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28945972 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28945901 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28945869 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28945851 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28945919 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28945928 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28945974 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28945882 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28945875 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28945936 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28945838 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28945871 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28945917 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28945897 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28945856 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28945883 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28945925 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28945860 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28945889 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28945911 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28945875 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10989542 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28945967 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28945917 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28945933 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28945871 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28945943 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28945916 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28945936 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28945911 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28945921 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10905654 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28945879 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28945938 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28945972 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28945957 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10496443 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28945828 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10603787 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28945971 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10882430 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28945923 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28945947 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28945950 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28945877 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10728708 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28945915 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28945840 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28945880 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28945928 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28945873 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28945940 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28945830 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28945838 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28945962 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28945931 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10771967 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28945868 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28945959 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10887083 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28945914 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28945964 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28945939 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10905652 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28945833 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28945965 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28945925 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10593922 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28945832 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28945927 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28945953 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28945837 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28945958 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10685777 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28945929 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28945912 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28945881 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28945945 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28945955 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28945913 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28945841 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10408676 - -
          Affymetrix Mouse Gene 1.0 ST Array 10408672 - -
          Affymetrix Mouse Gene 1.0 ST Array 10408667 - -
          Affymetrix Mouse Gene 1.0 ST Array 10408670 - -
          Affymetrix Mouse Gene 1.0 ST Array 10408674 - -
          Affymetrix Mouse Gene 1.0 ST Array 10408675 - -
          Affymetrix Mouse Gene 1.0 ST Array 10408673 - -
          Affymetrix Mouse Gene 1.0 ST Array 10408668 - -
          Affymetrix Mouse Gene 1.0 ST Array 10408669 - -
          Affymetrix Mouse Gene 1.0 ST Array 10408671 - -
          Affymetrix HT MG-430 PM Array Plate 1424792_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1443750_PM_s_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10408666 - -
          Mouse Genome 430 2.0 Array 1424792_PM_at - -
          Mouse Genome 430 2.0 Array 1424792_at - -
          Mouse Genome 430 2.0 Array 1443750_s_at - -
          Mouse Genome 430 2.0 Array 1443750_PM_s_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 208366_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 208366_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1300011263.mm.1 NM_145938.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300011263.mm.1 NM_145938.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1300011263.mm.1 TC1300001804.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300011266.mm.1 NM_145938.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300011266.mm.1 NM_145938.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1300011266.mm.1 TC1300001804.mm.1 MTA TC
          Mouse Expression Set 430 1424792_at - -
          Mouse Expression Set 430 1424792_PM_at - -
          Mouse Genome 430A 2.0 Array 1424792_PM_at - -
          Mouse Genome 430A 2.0 Array 1424792_at - -
          Affymetrix HT Mouse Genome 430A Array 1424792_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1424792_at - -
          Affymetrix HT Mouse Genome 430B Array 1443750_s_at - -
          Affymetrix HT Mouse Genome 430B Array 1443750_PM_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 493908 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 4610021 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004610021 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2669460 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2669460 - -
          mouse-6 v1.1 (Illumina) scl44096.6_563-S - -
          Illumina Sentrix mouse-6.v1 bead array 4610021 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl44096.6_563-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4610021 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2669460 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2669460 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline