Search
Search

Tspan8| Gene Symbol: Tspan8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001168679.1 | NP_001162150.1 | 6 - 7 | 625 | 63 | |
| NM_001168680.1 | NP_001162151.1 | 6 - 7 | 661 | 63 | ||
| NM_146010.2 | NP_666122.1 | 5 - 6 | 767 | 63 | ||
| GenBank mRNA | AK087696.1 | - | 6 - 7 | 701 | 63 | |
| BC025461.1 | - | 6 - 7 | 633 | 5344986 | 63 | |
| BC027777.1 | - | 6 - 7 | 636 | 5351814 | 63 | |
| BC029068.1 | - | 6 - 7 | 657 | 4947664 | 63 | |
| BC029808.1 | - | 5 - 6 | 766 | 4947699 | 63 | |
| BC034198.1 | - | 5 - 6 | 540 | 4986549 | 63 | |
| DV060937.1 | - | 5 - 6 | 600 | 63 | ||
| DV065030.1 | - | 6 - 7 | 803 | 63 | ||
Set Membership: |
|