Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00524927_m1
          See other APBB3 GE Assays ›
          Gene Symbol:
          Apbb3
          Gene Name:
          amyloid beta (A4) precursor protein-binding, family B, member 3
          Gene Aliases:
          Fe65l2, Rirl2, TR2S
          Chromosome Location:
          Chr.18: 36671154 - 36679366 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00524927_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Apbb3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146085.1 NP_666197.1 3 - 4 611 140
          GenBank mRNA AK141878.1 - 3 - 4 513 140
          AK171634.1 - 3 - 4 562 140
          BC024136.1 - 3 - 4 367 140
          BC024809.1 - 3 - 4 611 5357681 140

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1800001748.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Mus musculus Mouse Alzheimer's Disease 1700 Inventoried Mouse_Alzheimer Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Apbb3

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of transcription, DNA-templated
          negative regulation of apoptotic process
          beta-amyloid binding
          transcription factor binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31625255 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31625258 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31625256 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31625260 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31625257 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31625259 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2049582 BC024136.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2049582 BC024809.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2049582 AK171634.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2049582 NM_146085.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 972549 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 389010 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 819941 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 242280 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 165261 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P125778 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2049582 BC024136.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2049582 BC024809.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2049582 AK171634.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2049582 NM_146085.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P277321 BC024136.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P277321 BC024809.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P277321 AK171634.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P277321 AK141878.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P277321 NM_146085.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31625233 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31625245 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31625241 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31625255 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31625253 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31625248 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31625236 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31625240 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31625194 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31625197 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31625214 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31625224 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31625192 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31625223 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31625238 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31625196 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31625219 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31625215 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31625256 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31625221 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31625193 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31625226 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31625229 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31625239 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31625243 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31625247 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31625260 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31625252 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31625257 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31625237 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31625235 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31625234 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P125778 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08966807 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08966791 - -
          Whole Mouse Genome G4122A A_51_P277321 BC024136.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P277321 BC024809.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P277321 AK171634.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P277321 AK141878.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P277321 NM_146085.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P114159 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P125778 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13738773 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13738774 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13738784 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13738779 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31625258 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31625255 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31625259 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31625260 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31625257 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31625256 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625146 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625152 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625165 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625135 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625147 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625322 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625127 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625169 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625133 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625163 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625141 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625137 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625173 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625125 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625268 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625332 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625158 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625164 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625129 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625175 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625131 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625150 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625128 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625176 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625275 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625121 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625123 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625329 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625262 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625161 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625266 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625124 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625155 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625261 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625274 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625325 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625118 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625260 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625138 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625264 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625166 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625171 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625167 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13895734 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31625274 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31625268 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31625264 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31625260 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31625266 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31625289 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31625262 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31625261 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14091979 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31625293 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31625275 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31625271 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10458357 - -
          Affymetrix Mouse Gene 1.0 ST Array 10458363 - -
          Affymetrix Mouse Gene 1.0 ST Array 10458365 - -
          Affymetrix Mouse Gene 1.0 ST Array 10458361 - -
          Affymetrix Mouse Gene 1.0 ST Array 10458366 - -
          Affymetrix Mouse Gene 1.0 ST Array 10458360 - -
          Affymetrix Mouse Gene 1.0 ST Array 10458364 - -
          Affymetrix Mouse Gene 1.0 ST Array 10458356 - -
          Affymetrix Mouse Gene 1.0 ST Array 10458367 - -
          Affymetrix Mouse Gene 1.0 ST Array 10458359 - -
          Affymetrix Mouse Gene 1.0 ST Array 10458362 - -
          Affymetrix Mouse Gene 1.0 ST Array 10458358 - -
          Affymetrix HT MG-430 PM Array Plate 1424457_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10458355 - -
          Mouse Genome 430 2.0 Array 1424457_at - -
          Mouse Genome 430 2.0 Array 1424457_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 225372_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 225372_at - -
          Affymetrix Mouse Clariom S Array PSR1800008817.mm.1 NM_146085.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800008817.mm.1 NM_146085.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800008817.mm.1 TC1800001748.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800008818.mm.1 NM_146085.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800008818.mm.1 NM_146085.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800008818.mm.1 TC1800001748.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800008815.mm.1 NM_146085.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800008815.mm.1 NM_146085.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800008815.mm.1 TC1800001748.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800008819.mm.1 NM_146085.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800008819.mm.1 NM_146085.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800008819.mm.1 TC1800001748.mm.1 MTA TC
          Mouse Expression Set 430 1424457_at - -
          Mouse Expression Set 430 1424457_PM_at - -
          Mouse Genome 430A 2.0 Array 1424457_PM_at - -
          Mouse Genome 430A 2.0 Array 1424457_at - -
          Affymetrix HT Mouse Genome 430A Array 1424457_at - -
          Affymetrix HT Mouse Genome 430A Array 1424457_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 722977 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 6020168 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006020168 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1223271 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1223271 - -
          mouse-6 v1.1 (Illumina) scl51509.11.1_166-S - -
          Illumina Sentrix mouse-6.v1 bead array 6020168 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl51509.11.1_166-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6020168 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1223271 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1223271 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl51509.11.1_166-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline