Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00524986_m1
          See other UBXN1 GE Assays ›
          Gene Symbol:
          Ubxn1
          Gene Name:
          UBX domain protein 1
          Gene Aliases:
          2B28, 4930455J02Rik, D19Ertd721e, T25529
          Chromosome Location:
          Chr.19: 8871510 - 8875664 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00524986_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ubxn1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146093.1 NP_666205.1 5 - 6 340 76
          GenBank mRNA AK015468.1 - 4 - 5 392 76
          AK150442.1 - 4 - 5 343 76
          AK159708.1 - 4 - 5 316 76
          AK172428.1 - 2 - 3 694 76
          BC006701.1 - 5 - 6 340 3583916 76

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1900000162.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ubxn1

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of protein ubiquitination
          negative regulation of proteasomal ubiquitin-dependent protein catabolic process
          polyubiquitin binding
          ubiquitin protein ligase binding
          ubiquitin binding
          ATPase binding
          K6-linked polyubiquitin binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P568805 AK172428.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P568805 BC006701.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P568805 NM_146093.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P568805 AK015468.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P568805 AK150442.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P568805 AK172428.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P568805 BC006701.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P568805 NM_146093.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P568805 AK015468.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P568805 AK150442.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 968189 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 254171 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 109269 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P101276 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P568805 AK172428.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P568805 BC006701.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P568805 NM_146093.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P568805 AK015468.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P568805 AK150442.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P568805 AK172428.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P568805 BC006701.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P568805 NM_146093.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P568805 AK015468.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P568805 AK150442.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P249250 AK015468.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P568805 AK172428.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P568805 BC006701.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P568805 NM_146093.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P568805 AK015468.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P568805 AK150442.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P568805 AK172428.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P568805 BC006701.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P568805 NM_146093.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P568805 AK015468.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P568805 AK150442.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P119856 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14385062 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14327270 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14385065 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10461263 - -
          Affymetrix Mouse Gene 1.0 ST Array 10461267 - -
          Affymetrix Mouse Gene 1.0 ST Array 10461262 - -
          Affymetrix Mouse Gene 1.0 ST Array 10461265 - -
          Affymetrix Mouse Gene 1.0 ST Array 10461264 - -
          Affymetrix Mouse Gene 1.0 ST Array 10461259 - -
          Affymetrix Mouse Gene 1.0 ST Array 10461260 - -
          Affymetrix Mouse Gene 1.0 ST Array 10461258 - -
          Affymetrix Mouse Gene 1.0 ST Array 10461261 - -
          Affymetrix HT MG-430 PM Array Plate 1436342_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10461257 - -
          Mouse Genome 430 2.0 Array 1436342_a_at - -
          Mouse Genome 430 2.0 Array 1436342_PM_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 225896_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 225896_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1900002170.mm.1 NM_146093.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900002170.mm.1 NM_146093.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900002170.mm.1 TC1900000162.mm.1 MTA TC
          Mouse Expression Set 430 1436342_a_at - -
          Mouse Expression Set 430 1436342_PM_a_at - -
          Murine Genome U74v2 Set 97240_PM_g_at - -
          Murine Genome U74v2 Set 97240_g_at - -
          Murine Genome U74v2 Set 97239_PM_at - -
          Murine Genome U74v2 Set 97241_at - -
          Murine Genome U74v2 Set 97239_at - -
          Murine Genome U74v2 Set 97241_PM_at - -
          Mouse Genome 430A 2.0 Array 1436342_a_at - -
          Mouse Genome 430A 2.0 Array 1436342_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1436342_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1436342_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 310294 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 780601 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000780601 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2589583 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2589583 - -
          mouse-6 v1.1 (Illumina) scl53400.8.1_51-S - -
          Illumina Sentrix mouse-6.v1 bead array 780601 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl53400.8.1_51-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 780601 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2589583 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2589583 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl53400.8.1_51-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline