Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00525080_m1
          See other AAMP GE Assays ›
          Gene Symbol:
          Aamp
          Gene Name:
          angio-associated migratory protein
          Gene Aliases:
          AU040907, Aamp-rs
          Chromosome Location:
          Chr.1: 74279840 - 74284738 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00525080_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Aamp
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001190444.1 NP_001177373.1 2 - 3 397 61
          NM_146110.3 NP_666222.2 2 - 3 394 61
          GenBank mRNA AK146470.1 - 2 - 3 337 61
          AK151820.1 - 2 - 3 306 61
          AK162750.1 - 2 - 3 322 61
          AK167621.1 - 2 - 3 318 61
          AK212494.1 - 1 - 2 38 61
          BC031795.1 - 1 - 2 86 3965387 61

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0100002582.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Aamp

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20347696 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20347698 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20347694 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20347693 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P624362 BC080766.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P624362 BC031795.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P624362 AK167621.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P624362 AK162750.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P624362 AK146470.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P624362 BC080766.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P624362 BC031795.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P624362 AK167621.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P624362 AK162750.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P624362 AK146470.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 876773 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 427874 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 59570 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 398026 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 105361 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P161724 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P624362 BC080766.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P624362 BC031795.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P624362 AK167621.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P624362 AK162750.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P624362 AK146470.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P624362 BC080766.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P624362 BC031795.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P624362 AK167621.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P624362 AK162750.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P624362 AK146470.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P247591 BC080766.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P247591 BC031795.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P247591 AK167621.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P247591 AK185087.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P247591 AK187699.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20347676 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20347673 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20347692 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20347689 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20347658 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20347654 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20347663 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20347667 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20347672 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20347659 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20347661 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20347698 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20347653 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20347694 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20347696 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20347685 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20347671 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20347680 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20347681 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20347656 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20347660 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20347684 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20347679 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20347675 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20347664 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20347678 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20347682 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P161724 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04147367 - -
          Whole Mouse Genome G4122A A_51_P247591 BC080766.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P247591 BC031795.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P247591 AK167621.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P247591 AK185087.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P247591 AK187699.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P624362 BC080766.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P624362 BC031795.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P624362 AK167621.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P624362 AK162750.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P624362 AK146470.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P624362 BC080766.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P624362 BC031795.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P624362 AK167621.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P624362 AK162750.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P624362 AK146470.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P120973 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P137159 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P161724 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00323003 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00323011 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00323007 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00323000 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00323009 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20347696 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20347693 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20347698 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20347694 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20347634 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20347615 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20347626 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20347606 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20347686 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20347638 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20347678 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20347618 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20347681 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20347611 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20347675 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20347668 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20347617 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20347663 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20347689 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20347672 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20347628 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20347654 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20347676 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20347671 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20347609 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20347640 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20347653 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20347650 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20347595 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20347623 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20347621 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20347694 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20347603 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20347647 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20347685 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20347642 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20347664 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20347682 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20347698 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20347660 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20347632 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20347696 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20347604 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20347692 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20347656 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20347629 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20347613 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20347644 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20347641 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20347607 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20347673 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20347605 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20347667 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20347624 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20347658 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00388482 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20347668 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00388499 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00388483 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00388494 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20347673 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20347654 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00388467 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20347686 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00323011 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00388466 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00388506 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20347664 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20347685 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20347663 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00388488 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00388457 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20347682 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20347694 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00388496 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20347696 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20347667 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00388475 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20347692 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20347675 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10355555 - -
          Affymetrix Mouse Gene 1.0 ST Array 10355561 - -
          Affymetrix Mouse Gene 1.0 ST Array 10355560 - -
          Affymetrix Mouse Gene 1.0 ST Array 10355566 - -
          Affymetrix Mouse Gene 1.0 ST Array 10355565 - -
          Affymetrix Mouse Gene 1.0 ST Array 10355562 - -
          Affymetrix Mouse Gene 1.0 ST Array 10355557 - -
          Affymetrix Mouse Gene 1.0 ST Array 10355559 - -
          Affymetrix Mouse Gene 1.0 ST Array 10355556 - -
          Affymetrix Mouse Gene 1.0 ST Array 10355563 - -
          Affymetrix Mouse Gene 1.0 ST Array 10355564 - -
          Affymetrix Mouse Gene 1.0 ST Array 10355558 - -
          Affymetrix HT MG-430 PM Array Plate 1454754_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10355554 - -
          Mouse Genome 430 2.0 Array 1454754_PM_a_at - -
          Mouse Genome 430 2.0 Array 1454754_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 227290_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 227290_at - -
          Affymetrix Mouse Clariom S Array PSR0100020787.mm.1 NM_001190444.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100020787.mm.1 NM_001190444.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100020787.mm.1 NM_146110.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100020787.mm.1 NM_146110.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0100020787.mm.1 TC0100002582.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100020788.mm.1 NM_001190444.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100020788.mm.1 NM_001190444.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100020788.mm.1 NM_146110.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100020788.mm.1 NM_146110.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0100020788.mm.1 TC0100002582.mm.1 MTA TC
          Mouse Expression Set 430 1454754_a_at - -
          Mouse Expression Set 430 1454754_PM_a_at - -
          Murine Genome U74v2 Set 98064_PM_at - -
          Murine Genome U74v2 Set 98064_at - -
          Mouse Genome 430A 2.0 Array 1454754_a_at - -
          Mouse Genome 430A 2.0 Array 1454754_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1454754_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1454754_PM_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 327280 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2640059 - -
          Illumina mouse-6 v1.0 expression beadchip 2640059 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002640059 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2699416 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2699416 - -
          mouse-6 v1.1 (Illumina) scl0227290.1_240-S - -
          Illumina Sentrix mouse-6.v1 bead array 2640059 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0227290.1_240-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2640059 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1234981 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1234981 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0227290.1_240-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline