Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Gene Symbol: Nrd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146150.2 | NP_666262.2 | 5 - 6 | 999 | 60 | |
GenBank mRNA | AK031548.1 | - | 5 - 6 | 999 | 60 | |
AK040377.1 | - | 4 - 5 | 1230 | 60 | ||
AK166470.1 | - | 5 - 6 | 1129 | 60 | ||
BC023786.1 | - | 4 - 5 | 377 | 60 | ||
BC026832.1 | - | 5 - 6 | 1049 | 4486176 | 60 | |
BC036128.1 | - | 5 - 6 | 1117 | 4971791 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Mus musculus Probe spans exons |