Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00525375_m1
          See other ZFP418 GE Assays ›
          Gene Symbol:
          Zfp418
          Gene Name:
          zinc finger protein 418
          Gene Aliases:
          A230102I05Rik
          Chromosome Location:
          Chr.7: 7171353 - 7183560 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00525375_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zfp418
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146179.2 NP_666291.2 3 - 4 365 84
          GenBank mRNA AK039147.1 - 3 - 4 327 84
          AK054428.1 - 3 - 4 363 84
          BC024761.1 - 3 - 4 345 5363178 84
          BC026490.1 - 3 - 4 341 4504776 84

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700000101.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Zfp418

          Panther Classification:

          Molecular Function -

          C2H2 zinc finger transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P382512 BC024761.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P382512 BC026490.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P382512 AK039147.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P382512 AK054428.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P382512 BB671687.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P382512 BC024761.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P382512 BC026490.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P382512 AK039147.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P382512 AK054428.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P382512 BB671687.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 810327 AK047500.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 810327 XM_001474673.1 refseq_mrna_mouse_XM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 850209 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 518186 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 593500 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 779951 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 948400 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 678612 AK036330.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 299266 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 80050 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P382512 BC024761.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P382512 BC026490.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P382512 AK039147.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P382512 AK054428.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P382512 BB671687.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P382512 BC024761.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P382512 BC026490.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P382512 AK039147.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P382512 AK054428.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P382512 BB671687.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P382512 BC024761.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P382512 BC026490.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P382512 AK039147.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P382512 AK054428.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P382512 BB671687.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P382512 BC024761.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P382512 BC026490.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P382512 AK039147.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P382512 AK054428.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P382512 BB671687.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24958305 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24958325 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24958322 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24958315 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24958310 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24958326 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24958304 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24958320 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24958300 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24958303 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24958324 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24958307 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24958318 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24958314 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24958323 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24958313 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24958308 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24958298 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24958312 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01478053 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01478040 - -
          Whole Mouse Genome G4122A A_51_P382512 BC024761.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P382512 BC026490.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P382512 AK039147.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P382512 AK054428.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P382512 BB671687.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P382512 BC024761.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P382512 BC026490.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P382512 AK039147.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P382512 AK054428.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P382512 BB671687.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P107406 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P122770 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P01478053 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06008268 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06008274 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06414696 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06414693 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06008273 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24958360 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24958389 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24958300 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24958320 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24958308 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24958384 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24958298 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24958353 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24958390 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24958370 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24958350 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24958358 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24958363 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24958325 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24958373 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24958307 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24958314 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24958326 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24958375 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24958318 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24958322 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24958368 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24958337 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24958331 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24958366 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24958303 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24958334 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24958388 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24958347 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24958378 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24958362 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24958310 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24958367 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24958333 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24958312 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24958391 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24958305 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24958348 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24958339 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24958329 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24958379 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24958382 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24958324 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24958327 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24958342 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24958315 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24958387 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24958355 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24958345 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24958301 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06417371 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24958322 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06254891 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06444807 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24958318 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24958329 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24958319 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24958315 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24958333 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24958312 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24958314 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24958330 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24958327 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24958337 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24958344 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24958341 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06254895 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24958305 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24958307 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24958313 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24958303 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06417370 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24958310 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24958320 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05893369 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06102302 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24958308 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24958339 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06254892 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24958298 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10549900 - -
          Affymetrix Mouse Gene 1.0 ST Array 10549901 - -
          Affymetrix Mouse Gene 1.0 ST Array 10549903 - -
          Affymetrix Mouse Gene 1.0 ST Array 10549902 - -
          Affymetrix HT MG-430 PM Array Plate 1424974_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10549899 - -
          Mouse Genome 430 2.0 Array 1424974_at - -
          Mouse Genome 430 2.0 Array 1424974_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 232854_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 232854_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0700000965.mm.1 TC0700000101.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700000965.mm.1 NM_146179.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700000965.mm.1 NM_146179.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700000967.mm.1 TC0700000101.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700000967.mm.1 NM_146179.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700000967.mm.1 NM_146179.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700000963.mm.1 TC0700000101.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700000963.mm.1 NM_146179.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700000963.mm.1 NM_146179.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700000968.mm.1 TC0700000101.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700000968.mm.1 NM_146179.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700000968.mm.1 NM_146179.2 Refseq
          Mouse Expression Set 430 1424974_at - -
          Mouse Expression Set 430 1424974_PM_at - -
          Mouse Genome 430A 2.0 Array 1424974_at - -
          Mouse Genome 430A 2.0 Array 1424974_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1424974_at - -
          Affymetrix HT Mouse Genome 430A Array 1424974_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 335963 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6770484 - -
          Illumina mouse-6 v1.0 expression beadchip 6770484 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006770484 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1251225 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1251225 - -
          mouse-6 v1.1 (Illumina) scl33088.3_510-S - -
          Illumina Sentrix mouse-6.v1 bead array 6770484 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl33088.3_510-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6770484 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1251225 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1251225 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl33088.3_510-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline