| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1979563 |
NM_146186.3 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1979563 |
AK154592.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1979563 |
BC054747.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1979563 |
AK006173.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2105808 |
BC057041.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2105808 |
NM_146186.3 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2105808 |
BC054747.1 |
mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
737623 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
948039 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
384952 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
232604 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
306300 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
94509 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
491331 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
349116 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
506459 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
19709 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
260851 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
822484 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
931690 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
863375 |
AF253052.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
863375 |
BC040395.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
863375 |
NM_021899.2 |
refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
55583 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
382708 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
68342 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
488867 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
563070 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
606217 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
53019 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
608693 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
358996 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
150417 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
79345 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
2438 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
153921 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P126317 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P173116 |
- |
- |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1979563 |
NM_146186.3 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1979563 |
AK154592.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1979563 |
BC054747.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1979563 |
AK006173.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2105808 |
BC057041.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2105808 |
NM_146186.3 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2105808 |
BC054747.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P336114 |
BC057041.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P336114 |
NM_146186.3 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P336114 |
AK154592.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P336114 |
BC026444.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P336114 |
BC054747.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P420005 |
BC057041.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P420005 |
NM_146186.3 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P420005 |
AK154592.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P420005 |
AK209918.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P420005 |
BC026444.1 |
mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P135050 |
- |
- |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P108959 |
- |
- |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P113385 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25030679 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06503797 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P05993794 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25030676 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25030645 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06104237 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25030656 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06435880 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25030638 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25030571 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25030590 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P05914521 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25030625 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25030626 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06503792 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06435882 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P05968169 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25030624 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06032935 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25030657 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25030663 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06161943 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25030587 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25030654 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25030589 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25030741 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25030633 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P05954008 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25030584 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25030631 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25030618 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25030583 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06104238 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25030600 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25030640 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25030651 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25030599 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25030601 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P05968173 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06271847 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06104239 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25030573 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P05917817 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06271816 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25030602 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25030639 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06232171 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25030672 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25030710 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06538027 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25030659 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P05914527 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25030623 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06538026 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25030596 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25030668 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25030636 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P05932045 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06271843 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25030700 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06150816 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25030661 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25030634 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25030706 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06232172 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25030592 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25030658 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25030641 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P05914530 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06243215 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25030682 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25030642 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06376899 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25030711 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25030612 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06052203 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25030630 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25030574 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25030703 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25030616 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P05911697 |
- |
- |