Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00525519_m1
          See other SLC22A26 GE Assays ›
          Gene Symbol:
          Slc22a26
          Gene Name:
          solute carrier family 22 (organic cation transporter), member 26
          Gene Aliases:
          -
          Chromosome Location:
          Chr.19: 7781980 - 7802667 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00525519_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Slc22a26
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146232.1 NP_666344.1 8 - 9 1615 115
          GenBank mRNA AJ132857.1 - 1523 115
          BC014805.1 - 8 - 9 1615 4242162 115

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1900001056.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Slc22a26

          Panther Classification:

          Molecular Function -

          secondary carrier transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2020428 BC014805.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2020428 NM_146232.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2020428 AJ132857.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 124568 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 418227 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 448477 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 604800 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 222110 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 677190 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 927387 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 132818 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 971657 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 415503 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 450120 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 38327 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 141893 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 167420 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2020428 BC014805.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2020428 NM_146232.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2020428 AJ132857.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31941217 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31941207 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31941224 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31941229 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31941227 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31941223 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31941208 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31941228 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31941219 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31941216 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31941206 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31941218 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31941221 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31941215 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09099442 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03546702 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09099423 - -
          Whole Mouse Genome G4122A A_52_P460393 BC014805.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P460393 NM_146232.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P460393 AJ132857.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P299527 AJ132857.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P134778 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P101000 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P03546702 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P09099423 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14212201 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14212203 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31941128 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31941224 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31941177 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31941202 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31941155 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31941145 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31941216 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31941223 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31941168 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31941173 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31941141 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31941172 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31941204 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31941139 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31941228 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31941143 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31941138 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31941147 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31941135 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31941158 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31941176 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31941215 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31941134 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31941198 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31941146 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31941162 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31941218 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31941126 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31941137 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31941136 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31941156 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14306390 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31941123 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31941125 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31941128 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31941199 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31941190 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31941202 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31941129 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31941208 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31941204 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31941229 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31941224 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31941198 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14427809 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31941221 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31941218 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31941188 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31941126 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31941200 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31941217 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31941216 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31941223 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31941215 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31941228 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10465733 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465730 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465732 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465728 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465731 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465729 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465727 - -
          Affymetrix HT MG-430 PM Array Plate 1425752_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1425751_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10465726 - -
          Mouse Genome 430 2.0 Array 1425751_PM_at - -
          Mouse Genome 430 2.0 Array 1425752_at - -
          Mouse Genome 430 2.0 Array 1425751_at - -
          Mouse Genome 430 2.0 Array 1425752_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 236149_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 236149_at - -
          Affymetrix Mouse Clariom S Array PSR1900009857.mm.1 NM_146232.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900009857.mm.1 NM_146232.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900009857.mm.1 TC1900001056.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900009856.mm.1 NM_146232.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900009856.mm.1 NM_146232.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900009856.mm.1 TC1900001056.mm.1 MTA TC
          Mouse Expression Set 430 1425752_PM_at - -
          Mouse Expression Set 430 1425751_at - -
          Mouse Expression Set 430 1425752_at - -
          Mouse Expression Set 430 1425751_PM_at - -
          Mouse Genome 430A 2.0 Array 1425751_PM_at - -
          Mouse Genome 430A 2.0 Array 1425752_PM_at - -
          Mouse Genome 430A 2.0 Array 1425752_at - -
          Mouse Genome 430A 2.0 Array 1425751_at - -
          Affymetrix HT Mouse Genome 430A Array 1425752_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1425751_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1425751_at - -
          Affymetrix HT Mouse Genome 430A Array 1425752_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 778174 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 3130132 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003130132 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2628854 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2628854 - -
          mouse-6 v1.1 (Illumina) scl0236149.1_28-S - -
          Illumina Sentrix mouse-6.v1 bead array 3130132 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0236149.1_28-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3130132 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2959409 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2959409 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline