Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00525543_m1
          See other LRIT1 GE Assays ›
          Gene Symbol:
          Lrit1
          Gene Name:
          leucine-rich repeat, immunoglobulin-like and transmembrane domains 1
          Gene Aliases:
          BC032270, Lrrc21
          Chromosome Location:
          Chr.14: 37054830 - 37064938 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00525543_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lrit1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146245.2 NP_666357.2 1 - 2 218 75
          GenBank mRNA AK044631.1 - 1 - 2 219 75
          BC032270.1 - 1 - 2 224 5401567 75

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1400000392.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Lrit1

          Panther Classification:

          Molecular Function -

          immunoglobulin superfamily cell adhesion molecule

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1979295 NM_146245.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1979295 AK044631.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1979295 BC032270.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 510253 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 958258 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 176923 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 143097 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 401164 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 89649 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P168790 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P173717 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1979295 NM_146245.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1979295 AK044631.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1979295 BC032270.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29542961 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29542968 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29543017 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29542958 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29542954 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29542965 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29542966 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29543016 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29543011 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29543012 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29543020 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29543006 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29543010 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29542953 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29543008 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29542956 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29542960 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29543018 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29542971 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29543014 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29542963 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P168790 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P173717 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02834838 - -
          Whole Mouse Genome G4122A A_51_P145020 NM_146245.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P145020 AK044631.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P145020 BC032270.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P132225 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P168790 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P173717 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11737594 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11737591 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29542999 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29542930 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29543057 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29542997 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29542968 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29542990 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29542970 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29542977 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29543049 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29542939 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29542931 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29542904 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29543054 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29542923 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29542973 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29543004 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29542947 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29543001 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29542913 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29542941 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29543075 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29542974 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29543032 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29543025 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29543023 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29542950 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29543038 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29542911 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29542994 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29543064 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29543060 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29543087 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29542985 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29542927 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29543014 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29543063 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29543031 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29543016 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29542989 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29542945 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29543012 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29543011 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29542995 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29543040 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29542916 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29543008 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29542992 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29543091 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29543035 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29542993 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29542991 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29543047 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29542917 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29543079 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29543070 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29542936 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29542953 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29543045 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29543068 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29543072 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29542925 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29542899 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29543073 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29543043 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29542951 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29543044 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29543061 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29543089 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29542956 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29542919 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29543003 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29543088 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29542926 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29542965 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29543007 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29542966 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29542908 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29543020 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29543034 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29542954 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29543052 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29542910 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29543085 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29543000 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29542934 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29543006 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29543059 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29543037 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29542914 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29542921 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29542946 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29542901 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29542907 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29543041 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29542986 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29543083 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29543028 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29542963 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29543066 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29542958 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29543022 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29543009 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29542976 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29543010 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11292112 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29543007 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29542939 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29542977 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29542974 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11292104 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29542956 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29542966 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11661940 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29542946 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11737580 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29542960 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11168751 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11702072 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29542954 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29543008 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29542994 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29542961 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11292101 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29543020 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29542951 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29542963 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29542997 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11702059 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29542973 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29543018 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11737590 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29542992 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11661948 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11409070 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29542958 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11737591 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29542988 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29543003 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29543014 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11702057 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29543016 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29543004 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29542980 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29542950 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29542993 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29543009 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11292106 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11737581 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29542991 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11652179 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29543011 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29543012 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29542965 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11661947 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29542941 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10414167 - -
          Affymetrix Mouse Gene 1.0 ST Array 10414165 - -
          Affymetrix Mouse Gene 1.0 ST Array 10414166 - -
          Affymetrix Mouse Gene 1.0 ST Array 10414164 - -
          Affymetrix HT MG-430 PM Array Plate 1441110_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10414163 - -
          Mouse Genome 430 2.0 Array 1441110_PM_at - -
          Mouse Genome 430 2.0 Array 1441110_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 239037_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 239037_at - -
          Affymetrix Mouse Clariom S Array PSR1400003489.mm.1 NM_146245.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400003489.mm.1 NM_146245.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1400003489.mm.1 TC1400000392.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400003488.mm.1 NM_146245.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400003488.mm.1 NM_146245.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1400003488.mm.1 TC1400000392.mm.1 MTA TC
          Mouse Expression Set 430 1441110_PM_at - -
          Mouse Expression Set 430 1441110_at - -
          Affymetrix HT Mouse Genome 430B Array 1441110_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1441110_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 460676 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 7040736 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0007040736 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2589757 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2589757 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2525924 - -
          mouse-6 v1.1 (Illumina) scl46440.4.1_161-S - -
          Illumina Sentrix mouse-6.v1 bead array 7040736 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl46440.4.1_161-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 7040736 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2589757 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2589757 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline