Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00525626_s1
          See other OLFR448 GE Assays ›
          Gene Symbol:
          Olfr448
          Gene Name:
          olfactory receptor 448
          Gene Aliases:
          MOR261-13
          Chromosome Location:
          Chr.6: 42896453 - 42897385 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00525626_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr448
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146273.1 NP_666385.1 1 449 57
          GenBank mRNA BC107336.1 - 1 441 57
          BC107337.1 - 1 449 40055003 57

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr448

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2104168 BC107337.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2104168 NM_146273.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2104168 BC107336.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P161552 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2104168 BC107337.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2104168 NM_146273.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2104168 BC107336.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P376384 BC107337.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P376384 NM_146273.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P376384 BC107336.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400628 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400616 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400610 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400623 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400620 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400609 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400604 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400607 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400602 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400619 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400624 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400605 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400630 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400626 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400612 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400617 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400621 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400614 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400606 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P161552 - -
          Whole Mouse Genome G4122A A_51_P376384 BC107337.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P376384 NM_146273.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P376384 BC107336.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P107371 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P161552 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05176923 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05176904 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05176900 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05176927 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400602 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400659 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400616 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400623 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400631 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400595 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400664 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400671 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400634 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400672 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400660 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400619 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400609 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400598 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400617 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400607 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400594 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400590 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400593 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400589 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400636 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400605 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400633 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400630 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400592 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400661 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400614 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400612 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400628 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400662 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400626 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400610 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400596 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400620 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400638 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400624 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400601 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400617 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400628 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05086031 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400607 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400632 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400589 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400633 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400592 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400619 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400621 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400612 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400644 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400610 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05382330 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400631 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400590 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400636 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05468206 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400614 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400605 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400620 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400606 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400602 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400623 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400630 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400638 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400626 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400616 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400634 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400596 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400624 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400595 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400642 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400601 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400600 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400598 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05176900 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10537804 - -
          Affymetrix Mouse Gene 1.1 ST Array 10537803 - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 808998 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 1690164 - -
          Illumina mouse-6 v1.0 expression beadchip 1690164 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001690164 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2645100 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2645100 - -
          mouse-6 v1.1 (Illumina) scl1727.1.1_48-S - -
          Illumina Sentrix mouse-6.v1 bead array 1690164 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl1727.1.1_48-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1690164 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2645100 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2645100 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl1727.1.1_48-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-mrft7:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0