Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00525652_s1
          See other OLFR1394 GE Assays ›
          Gene Symbol:
          Olfr1394
          Gene Name:
          olfactory receptor 1394
          Gene Aliases:
          MOR280-1
          Chromosome Location:
          Chr.11: 49160016 - 49160954 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00525652_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr1394
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146276.1 NP_666388.1 1 55 73
          GenBank mRNA BC127968.1 - 1 59 40135034 73

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100000498.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr1394

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2113954 NM_146276.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2113954 BC127968.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 236880 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P165518 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2113954 NM_146276.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2113954 BC127968.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P285462 NM_146276.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P285462 BC127968.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27781575 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27781576 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27781560 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27781552 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27781529 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27781553 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27781557 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27781586 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27781564 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27781595 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27781528 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27781590 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27781555 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27781556 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27781597 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27781602 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27781567 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27781580 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27781579 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27781565 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27781584 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27781572 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27781601 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27781583 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27781594 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27781554 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27781599 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27781592 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27781562 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27781570 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27781600 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P165518 - -
          Whole Mouse Genome G4122A A_51_P285462 NM_146276.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P285462 BC127968.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P104394 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P165518 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09016618 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09294449 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09294452 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09294454 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27781604 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27781557 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27781538 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27781528 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27781611 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27781606 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27781600 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27781531 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27781601 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27781563 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27781534 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27781576 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27781590 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27781575 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27781544 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27781555 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27781579 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27781536 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27781546 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27781580 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27781597 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27781537 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27781592 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27781560 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27781584 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27781586 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27781535 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27781552 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27781609 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27781532 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27781556 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27781583 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27781608 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27781565 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27781599 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27781553 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27781594 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27781550 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27781603 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27781602 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27781581 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27781533 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27781594 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27781556 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27781557 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27781533 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27781579 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09243218 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27781560 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27781546 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27781567 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27781553 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09618918 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27781606 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27781586 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27781576 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27781532 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27781537 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27781570 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09015476 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09106606 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09294443 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27781531 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27781611 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27781580 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27781583 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27781603 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27781544 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27781596 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27781599 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09015473 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27781562 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27781559 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27781550 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27781558 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27781540 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27781609 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27781565 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09083844 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09705829 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27781536 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27781555 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27781575 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09690400 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09294441 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27781541 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27781572 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27781526 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09035304 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09035307 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27781612 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27781581 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27781590 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27781549 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27781584 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27781534 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09243220 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27781528 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09294449 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27781530 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27781597 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27781608 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10375528 - -
          Affymetrix Mouse Gene 1.1 ST Array 10375527 - -
          Affymetrix Mouse Clariom S Array PSR1100003810.mm.1 NM_146276.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100003810.mm.1 NM_146276.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100003810.mm.1 TC1100000498.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 530624 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 360095 - -
          Illumina mouse-6 v1.0 expression beadchip 360095 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000360095 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2616214 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2616214 - -
          mouse-6 v1.1 (Illumina) scl14141.1.1_218-S - -
          Illumina Sentrix mouse-6.v1 bead array 360095 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl14141.1.1_218-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 360095 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2616214 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2616214 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl14141.1.1_218-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-6wvps:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0