Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00525665_s1
          See other OLFR1412 GE Assays ›
          Gene Symbol:
          Olfr1412
          Gene Name:
          olfactory receptor 1412
          Gene Aliases:
          MOR208-4
          Chromosome Location:
          Chr.1: 92588332 - 92589297 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00525665_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr1412
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146277.1 NP_666389.1 1 345 91
          GenBank mRNA BC125241.1 - 1 345 40044004 91

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0100000909.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr1412

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          G-protein coupled receptor activity
          olfactory receptor activity
          odorant binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P403728 NM_146277.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P403728 BC125241.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P403728 NM_146277.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P403728 BC125241.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P152007 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P403728 NM_146277.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P403728 BC125241.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P403728 NM_146277.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P403728 BC125241.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P403728 NM_146277.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P403728 BC125241.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P403728 NM_146277.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P403728 BC125241.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20449753 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20449758 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20449767 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20449760 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20449770 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20449766 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20449785 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20449773 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20449768 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20449769 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20449771 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20449755 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20449746 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20449772 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20449774 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20449749 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20449764 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20449752 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20449763 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20449783 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20449761 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20449777 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P152007 - -
          Whole Mouse Genome G4122A A_51_P403728 NM_146277.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P403728 BC125241.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P403728 NM_146277.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P403728 BC125241.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P110350 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P152007 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00033939 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00033927 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00033938 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00033943 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20449807 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20449769 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20449822 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20449758 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20449745 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20449825 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20449829 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20449821 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20449766 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20449828 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20449785 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20449755 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20449749 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20449809 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20449771 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20449810 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20449839 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20449838 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20449853 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20449827 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20449768 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20449774 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20449823 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20449752 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20449764 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20449840 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20449814 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20449761 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20449818 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20449753 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20449849 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20449851 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20449760 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20449783 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20449812 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00033933 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20449772 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00033954 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00033944 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00033945 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00033919 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00033922 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00033936 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00981505 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20449768 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20449752 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20449749 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00033928 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00033943 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20449755 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00033916 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20449761 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00033938 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20449746 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00033937 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00033948 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20449745 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20449767 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20449753 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10348638 - -
          Affymetrix Mouse Gene 1.1 ST Array 10348637 - -
          Affymetrix Mouse Clariom S Array PSR0100007663.mm.1 NM_146277.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100007663.mm.1 NM_146277.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100007663.mm.1 TC0100000909.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 681026 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 3190601 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003190601 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2699150 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2699150 - -
          mouse-6 v1.1 (Illumina) scl17641.1.21_43-S - -
          Illumina Sentrix mouse-6.v1 bead array 3190601 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl17641.1.21_43-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3190601 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2699148 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2699148 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-4lm45:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0