Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00525708_s1
          See other OLFR284 GE Assays ›
          Gene Symbol:
          Olfr284
          Gene Name:
          olfactory receptor 284
          Gene Aliases:
          MOR160-4
          Chromosome Location:
          Chr.15: 98340022 - 98340939 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00525708_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr284
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146281.1 NP_666393.1 1 748 67
          GenBank mRNA BC111907.1 - 1 737 67

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr284

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception
          detection of chemical stimulus involved in sensory perception of smell
          transmembrane signaling receptor activity
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2159114 BC111907.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2159114 NM_146281.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P174288 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2159114 BC111907.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2159114 NM_146281.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P505457 BC111907.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P505457 NM_146281.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30474194 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30474169 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30474217 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30474202 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30474178 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30474192 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30474219 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30474179 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30474170 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30474212 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30474174 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30474167 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30474171 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30474208 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30474195 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30474191 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30474189 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30474173 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30474201 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30474215 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30474196 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30474198 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30474168 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30474204 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30474186 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30474165 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30474177 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30474214 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30474187 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30474183 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30474188 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30474180 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30474190 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30474206 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30474210 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30474175 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30474200 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30474176 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30474181 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P174288 - -
          Whole Mouse Genome G4122A A_51_P505457 BC111907.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P505457 NM_146281.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P109206 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P113570 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P174288 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12324871 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12324868 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12324858 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12220764 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12220749 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12324864 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474215 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474222 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474192 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474217 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474226 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474137 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474191 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474223 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474195 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474168 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474202 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474169 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474144 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474149 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474212 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474183 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474210 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474157 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474161 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474151 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474173 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474152 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474206 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474140 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474131 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474171 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474208 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474204 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474196 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474128 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474188 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474162 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474220 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474124 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474176 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474184 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474186 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474187 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474179 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474141 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474123 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474178 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474214 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474181 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474174 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474224 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474139 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474142 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30474194 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12333356 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474199 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474181 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474206 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474210 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474190 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12220753 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474217 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474141 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474137 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12324871 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474184 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474188 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474162 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474175 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474155 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474170 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474204 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474149 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11917475 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474223 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474194 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474165 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474168 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474196 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474200 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11917474 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474220 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474139 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474183 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474214 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474229 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474222 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474224 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474142 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474171 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474147 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11922497 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12220750 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12212235 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474173 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474180 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474191 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474164 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11845455 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11917487 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12206647 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474208 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474161 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474202 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474179 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11930969 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474174 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474226 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474159 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474169 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12220764 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11922493 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474148 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12431305 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474157 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474176 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474151 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474144 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474152 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30474150 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10432153 - -
          Affymetrix Mouse Gene 1.1 ST Array 10432152 - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 655143 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 5550181 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005550181 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2755939 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2755939 - -
          mouse-6 v1.1 (Illumina) scl46771.1.1_93-S - -
          Illumina Sentrix mouse-6.v1 bead array 5550181 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl46771.1.1_93-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5550181 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2755939 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2755939 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline