Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00525875_s1
          See other OLFR745 GE Assays ›
          Gene Symbol:
          Olfr745
          Gene Name:
          olfactory receptor 745
          Gene Aliases:
          MOR106-11
          Chromosome Location:
          Chr.14: 50642195 - 50643369 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00525875_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr745
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146299.2 NP_666411.2 1 631 79
          GenBank mRNA BC132579.1 - 1 629 40130856 79
          BC132581.1 - 1 629 40130858 79

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1400000562.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr745

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P430414 BC132581.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P430414 BC132579.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P430414 NM_146299.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P430414 BC132581.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P430414 BC132579.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P430414 NM_146299.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P161471 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P178860 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P113375 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P430414 BC132581.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P430414 BC132579.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P430414 NM_146299.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P430414 BC132581.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P430414 BC132579.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P430414 NM_146299.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P430414 BC132581.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P430414 BC132579.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P430414 NM_146299.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P430414 BC132581.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P430414 BC132579.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P430414 NM_146299.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597915 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597905 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597899 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597894 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597913 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597921 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597919 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597898 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597908 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597917 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597904 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597903 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597910 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597895 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597900 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597907 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597912 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597922 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P113375 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P161471 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P178860 - -
          Whole Mouse Genome G4122A A_51_P430414 BC132581.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P430414 BC132579.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P430414 NM_146299.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P430414 BC132581.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P430414 BC132579.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P430414 NM_146299.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P111312 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P113375 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P161471 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P178860 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11703679 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11755992 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11703672 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11703670 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597921 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597967 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597917 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597968 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597919 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597904 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597907 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597959 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597894 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597910 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597915 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597961 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597900 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597898 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597905 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597970 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597908 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597913 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597903 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597896 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597899 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597915 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597913 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597904 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11703669 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597910 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11755992 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597894 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597903 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11703670 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597908 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597922 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597898 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597919 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597905 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597900 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11703536 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597921 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11703673 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597907 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597899 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10414490 - -
          Affymetrix Mouse Gene 1.1 ST Array 10414489 - -
          Affymetrix Mouse Clariom S Array PSR1400004435.mm.1 NM_146299.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400004435.mm.1 NM_146299.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1400004435.mm.1 TC1400000562.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 772246 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 2650671 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002650671 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2607889 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2607889 - -
          mouse-6 v1.1 (Illumina) scl2751.1.1_270-S - -
          Illumina Sentrix mouse-6.v1 bead array 2650671 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl2751.1.1_270-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2650671 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2607889 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2607889 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-mrft7:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0