Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00525884_s1
          See other OLFR827 GE Assays ›
          Gene Symbol:
          Olfr827
          Gene Name:
          olfactory receptor 827
          Gene Aliases:
          MOR210-5
          Chromosome Location:
          Chr.10: 130210160 - 130211128 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00525884_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr827
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146300.1 NP_666412.1 1 456 63
          GenBank mRNA BC117109.1 - 1 477 40126428 63
          BC117111.1 - 1 477 40126430 63

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1000003206.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr827

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          G-protein coupled receptor activity
          olfactory receptor activity
          odorant binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2080855 NM_146300.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2080855 BC117109.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2080855 BC117111.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 400428 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P139835 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2080855 NM_146300.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2080855 BC117109.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2080855 BC117111.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P208857 NM_146300.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P208857 BC117109.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P208857 BC117111.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27548757 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27548759 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27548766 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27548740 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27548790 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27548773 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27548778 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27548733 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27548779 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27548771 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27548765 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27548738 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27548743 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27548768 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27548736 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27548784 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27548755 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27548734 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27548753 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27548787 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27548750 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27548776 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27548762 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27548764 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27548774 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27548786 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27548781 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27548742 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27548791 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27548754 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27548745 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27548780 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27548747 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27548775 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27548761 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P139835 - -
          Whole Mouse Genome G4122A A_51_P208857 NM_146300.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P208857 BC117109.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P208857 BC117111.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P132574 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P139835 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08555202 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08839869 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08555211 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08839867 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08555212 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548766 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548712 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548795 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548773 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548781 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548743 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548709 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548729 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548790 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548724 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548720 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548722 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548765 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548780 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548771 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548797 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548761 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548755 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548719 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548731 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548738 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548802 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548759 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548716 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548742 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548723 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548787 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548706 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548796 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548791 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548799 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548745 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548793 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548768 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548734 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548715 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548726 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548757 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548800 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548786 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548776 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548754 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548762 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548750 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548747 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548778 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548764 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548784 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548736 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27548740 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548745 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548771 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548795 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548743 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548798 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548799 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548784 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548790 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548786 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548738 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548742 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548774 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08536777 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548732 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548709 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548778 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548757 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548780 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548791 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548747 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548723 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548729 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548781 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548724 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548762 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548787 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548731 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548734 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548736 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08392827 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548726 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08536772 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548779 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548800 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08709561 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548712 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548764 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548711 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548719 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548720 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08555210 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548768 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548802 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548716 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548715 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548754 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548776 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548755 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08695677 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548740 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548797 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548750 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548759 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548773 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548717 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548722 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27548761 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10373679 - -
          Affymetrix Mouse Gene 1.1 ST Array 10373678 - -
          Affymetrix Mouse Clariom S Array PSR1000023991.mm.1 NM_146300.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000023991.mm.1 NM_146300.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000023991.mm.1 TC1000003206.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 463406 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 5360671 - -
          Illumina mouse-6 v1.0 expression beadchip 5360671 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005360671 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1230944 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1230944 - -
          mouse-6 v1.1 (Illumina) scl5151.1.1_53-S - -
          Illumina Sentrix mouse-6.v1 bead array 5360671 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl5151.1.1_53-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5360671 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1230944 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1230944 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-6wvps:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0