Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00525903_s1
          See other OLFR1449 GE Assays ›
          Gene Symbol:
          Olfr1449
          Gene Name:
          olfactory receptor 1449
          Gene Aliases:
          MOR202-34
          Chromosome Location:
          Chr.19: 12934740 - 12935684 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00525903_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr1449
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146303.1 NP_666415.1 1 454 76
          GenBank mRNA BC133705.1 - 1 458 40135405 76

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr1449

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          G-protein coupled receptor activity
          olfactory receptor activity
          odorant binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2180784 NM_146303.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2180784 BC133705.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2002978 NM_146303.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2002978 BC133705.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 75314 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P164025 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2002978 NM_146303.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2002978 BC133705.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2180784 NM_146303.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2180784 BC133705.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P494257 NM_146303.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P494257 BC133705.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31963115 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31963147 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31963108 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31963127 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31963109 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31963119 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31963131 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31963121 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31963120 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31963133 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31963132 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31963116 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31963143 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31963118 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31963150 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31963123 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31963113 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31963138 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31963145 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31963130 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31963111 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P164025 - -
          Whole Mouse Genome G4122A A_51_P494257 NM_146303.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P494257 BC133705.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P130633 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P164025 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14436081 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14436067 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14436082 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14436070 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14436072 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14436066 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31963096 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31963158 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31963097 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31963124 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31963138 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31963099 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31963154 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31963109 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31963111 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31963118 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31963141 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31963148 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31963143 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31963113 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31963145 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31963103 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31963121 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31963120 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31963150 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31963093 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31963108 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31963095 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31963106 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31963131 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31963127 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31963132 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31963145 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31963138 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31963136 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31963118 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31963124 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31963131 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31963130 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31963150 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31963102 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31963147 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31963110 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31963111 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14494120 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31963106 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14223582 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31963103 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31963120 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31963154 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31963113 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31963121 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14477670 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31963158 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31963108 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31963119 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31963116 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31963141 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14436072 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31963148 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31963123 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31963155 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31963162 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31963156 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31963132 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31963127 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10461785 - -
          Affymetrix Mouse Gene 1.1 ST Array 10461784 - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 581283 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 670195 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000670195 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1223962 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1223962 - -
          mouse-6 v1.1 (Illumina) scl6293.1.1_6-S - -
          Illumina Sentrix mouse-6.v1 bead array 670195 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl6293.1.1_6-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 670195 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1223962 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1223962 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-mrft7:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0