Search
Search

Olfr518| Gene Symbol: Olfr518 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146306.1 | NP_666418.1 | 1 | 52 | 83 | |
| GenBank mRNA | BC104128.1 | - | 1 | 52 | 83 | |
| BC104129.1 | - | 1 | 52 | 83 | ||
| BC114642.1 | - | 1 | 51 | 83 | ||
| BC115846.1 | - | 1 | 51 | 83 | ||
| BC127222.1 | - | 1 | 52 | 40044607 | 83 | |
| BC127223.1 | - | 1 | 52 | 40044609 | 83 | |
Set Membership: |
|