Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00526036_s1
          See other OLFR186 GE Assays ›
          Gene Symbol:
          Olfr186
          Gene Name:
          olfactory receptor 186
          Gene Aliases:
          MOR183-9
          Chromosome Location:
          Chr.16: 59026976 - 59027905 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00526036_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr186
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146321.1 NP_666433.1 1 665 86
          GenBank mRNA BC150713.1 - 1 687 9087624 86

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr186

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          signal transduction
          G-protein coupled receptor signaling pathway
          sensory perception of smell
          response to stimulus
          detection of chemical stimulus involved in sensory perception of smell
          signal transducer activity
          G-protein coupled receptor activity
          olfactory receptor activity
          odorant binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P390046 NM_146321.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P390046 BC150713.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P390046 NM_146321.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P390046 BC150713.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P152300 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P390046 NM_146321.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P390046 BC150713.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P390046 NM_146321.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P390046 BC150713.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P390046 NM_146321.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P390046 BC150713.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P390046 NM_146321.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P390046 BC150713.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30794038 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30794008 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30794031 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30794045 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30794057 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30794010 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30794033 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30794017 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30794040 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30794029 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30793999 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30794004 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30794027 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30794001 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30793996 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30794035 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30794028 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30794060 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30794024 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30793997 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30794014 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30794059 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30794041 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30794026 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30794043 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30794061 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30794018 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30794063 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30794020 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30794065 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30794016 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30793994 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30794006 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30794047 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30794037 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30794021 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P152300 - -
          Whole Mouse Genome G4122A A_51_P390046 NM_146321.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P390046 BC150713.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P390046 NM_146321.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P390046 BC150713.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P131877 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P152300 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12702347 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12603754 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12702327 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12702342 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12603758 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12702337 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794037 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794072 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794017 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794055 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30793984 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794020 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794075 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30793994 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794008 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794031 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794051 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794018 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794021 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794006 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794010 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794038 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794035 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794001 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794041 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30793979 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794029 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794065 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794076 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30793999 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794061 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794044 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794024 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30793978 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794058 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30793992 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794026 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794078 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794068 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794077 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30793987 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794053 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794033 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30793975 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794016 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30793971 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794004 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794063 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794073 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794027 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30793985 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794014 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794028 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794045 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794012 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30793969 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794060 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30793997 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794047 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30793994 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794010 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794026 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794061 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794065 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30793977 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30793975 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12744597 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12702341 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794021 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794027 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794037 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794038 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30793999 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794001 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30793969 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30793985 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30793982 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30793984 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794043 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794047 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794006 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794020 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794029 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794016 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794040 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30793997 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30793971 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794004 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794053 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794058 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794044 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794063 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794055 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12991701 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794035 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794033 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30793978 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30793968 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794041 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794054 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30793992 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30793970 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30793988 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794024 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794045 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794060 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12702344 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794051 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30793987 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30793979 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12702327 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794018 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794012 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794028 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794031 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10440150 - -
          Affymetrix Mouse Gene 1.1 ST Array 10440149 - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 415136 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4590520 - -
          Illumina mouse-6 v1.0 expression beadchip 4590520 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004590520 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2669909 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2669909 - -
          mouse-6 v1.1 (Illumina) scl479.1.1_5-S - -
          Illumina Sentrix mouse-6.v1 bead array 4590520 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl479.1.1_5-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4590520 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2669909 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2669909 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline