Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00526042_s1
          See other OLFR187 GE Assays ›
          Gene Symbol:
          Olfr187
          Gene Name:
          olfactory receptor 187
          Gene Aliases:
          MOR183-8
          Chromosome Location:
          Chr.16: 59035780 - 59039749 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00526042_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr187
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146322.2 NP_666434.2 2 359 108
          GenBank mRNA BC119304.1 - 1 247 8734053 108
          BC119306.1 - 1 250 8734055 108

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr187

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          signal transduction
          G-protein coupled receptor signaling pathway
          sensory perception of smell
          response to stimulus
          detection of chemical stimulus involved in sensory perception of smell
          signal transducer activity
          G-protein coupled receptor activity
          olfactory receptor activity
          odorant binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P386280 NM_146322.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P386280 NM_146322.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P146050 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P386280 NM_146322.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P386280 NM_146322.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P386280 NM_146322.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P386280 NM_146322.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30794090 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30794092 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30794100 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30794068 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30794099 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30794077 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30794083 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30794076 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30794087 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30794081 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30794091 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30794096 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30794085 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30794075 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30794070 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30794088 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30794097 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30794073 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30794093 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30794078 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P146050 - -
          Whole Mouse Genome G4122A A_51_P386280 NM_146322.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P386280 NM_146322.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P130568 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P146050 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12830544 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12830540 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12705880 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12830537 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12705878 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794010 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794026 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794008 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794083 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794001 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794018 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794076 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794093 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794012 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794108 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794087 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794021 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794110 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794035 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794027 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794017 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794020 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794099 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794041 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794038 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794004 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794029 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794097 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794075 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794037 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794091 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794107 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794073 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794081 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794014 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794088 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30793999 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794096 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30793997 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794078 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794085 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794077 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794033 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794112 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794016 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794111 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794072 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794090 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794024 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794068 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794031 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794006 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794028 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794093 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794091 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794083 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12830538 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794096 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794099 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794102 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794067 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794090 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794076 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794081 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794101 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794075 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794072 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794088 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794077 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794078 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794087 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794073 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794068 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794070 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12793884 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10440152 - -
          Affymetrix Mouse Gene 1.0 ST Array 10440153 - -
          Affymetrix Mouse Gene 1.1 ST Array 10440151 - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 740708 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 3940403 - -
          Illumina mouse-6 v1.0 expression beadchip 3940403 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003940403 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2599440 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2599440 - -
          mouse-6 v1.1 (Illumina) scl478.1.1_245-S - -
          Illumina Sentrix mouse-6.v1 bead array 3940403 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl478.1.1_245-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3940403 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2599440 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2599440 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-mrft7:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0