Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00526069_s1
          See other OLFR554 GE Assays ›
          Gene Symbol:
          Olfr554
          Gene Name:
          olfactory receptor 554
          Gene Aliases:
          MOR25-1
          Chromosome Location:
          Chr.7: 102640248 - 102641201 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00526069_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr554
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146325.2 NP_666437.2 1 253 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700001462.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr554

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1965255 NM_146325.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P131120 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1965255 NM_146325.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P109439 NM_146325.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P131120 - -
          Whole Mouse Genome G4122A A_51_P109439 NM_146325.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P129167 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P131120 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379831 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379838 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379864 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379841 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379845 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379909 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379835 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379867 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379844 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379826 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379854 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379822 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379859 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379906 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379871 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379850 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379832 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379911 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379904 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379840 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379913 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379855 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379823 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379834 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379870 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379833 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379868 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379902 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379860 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379857 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379828 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379858 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379862 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379847 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379839 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379864 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379842 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379828 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379850 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379868 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379826 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379829 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379839 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379902 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379881 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379878 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379835 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379831 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06662321 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06134984 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379871 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06525229 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379838 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379847 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06662339 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379822 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379870 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06662317 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379841 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379824 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06662345 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379862 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06525233 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379857 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06662343 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06386529 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379844 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379867 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379840 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10555728 - -
          Affymetrix Mouse Gene 1.1 ST Array 10555727 - -
          Affymetrix Mouse Clariom S Array PSR0700012687.mm.1 TC0700001462.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700012687.mm.1 NM_146325.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700012687.mm.1 NM_146325.2 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 410760 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 6420129 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006420129 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2590359 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2590359 - -
          mouse-6 v1.1 (Illumina) scl9413.1.1_57-S - -
          Illumina Sentrix mouse-6.v1 bead array 6420129 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl9413.1.1_57-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6420129 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2590359 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2590359 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl9413.1.1_57-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline