Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00526216_s1
          See other OLFR1495 GE Assays ›
          Gene Symbol:
          Olfr1495
          Gene Name:
          olfactory receptor 1495
          Gene Aliases:
          MOR266-9
          Chromosome Location:
          Chr.19: 13768344 - 13769303 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00526216_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr1495
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146344.1 NP_666456.1 1 49 71
          GenBank mRNA BC125566.1 - 1 86 40130081 71
          BC137879.1 - 1 84 8860898 71

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1900000283.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr1495

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          G-protein coupled receptor activity
          olfactory receptor activity
          odorant binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2106185 NM_146344.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2106185 BC125566.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2106185 BC137879.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P153039 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P171824 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P179928 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2106185 NM_146344.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2106185 BC125566.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2106185 BC137879.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P192639 NM_146344.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P192639 BC125566.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P192639 BC137879.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31965749 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31965727 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31965725 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31965708 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31965740 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31965751 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31965716 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31965744 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31965701 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31965696 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31965747 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31965736 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31965722 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31965754 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31965730 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31965702 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31965734 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31965710 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31965698 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31965717 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31965752 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31965719 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31965697 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31965738 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31965750 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31965721 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31965706 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31965743 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31965723 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31965707 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31965700 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31965732 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31965737 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31965715 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31965714 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31965704 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P153039 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P171824 - -
          Whole Mouse Genome G4122A A_51_P192639 NM_146344.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P192639 BC125566.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P192639 BC137879.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P127494 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P171824 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P153039 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14318736 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14318729 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14318734 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14318735 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14318728 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965730 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965723 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965736 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965698 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965704 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965764 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965697 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965762 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965701 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965768 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965696 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965737 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965752 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965744 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965717 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965727 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965749 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965721 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965706 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965760 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965719 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965715 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965747 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965708 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965771 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965734 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965758 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965722 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965738 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965702 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965725 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965766 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965700 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965714 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965750 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965753 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965743 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965710 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965715 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965738 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965725 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965723 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965740 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965749 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965714 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965701 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965737 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965697 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965744 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965710 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965716 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965719 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965742 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965696 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965702 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965752 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14318733 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965727 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14497290 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14270817 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965721 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965736 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965753 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965734 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965750 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965698 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965717 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965730 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965708 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14346247 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965747 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965754 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965695 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965743 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965704 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965722 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10461831 - -
          Affymetrix Mouse Gene 1.1 ST Array 10461830 - -
          Affymetrix Mouse Clariom S Array PSR1900003050.mm.1 TC1900000283.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 672720 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 670309 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000670309 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2609074 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2609074 - -
          mouse-6 v1.1 (Illumina) scl6277.1.1_295-S - -
          Illumina Sentrix mouse-6.v1 bead array 670309 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl6277.1.1_295-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 670309 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2609074 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2609074 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-jwmpg:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0