Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00526263_s1
          See other OLFR1121 GE Assays ›
          Gene Symbol:
          Olfr1121
          Gene Name:
          olfactory receptor 1121
          Gene Aliases:
          MOR264-18
          Chromosome Location:
          Chr.2: 87371534 - 87372478 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00526263_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr1121
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146348.2 NP_666460.2 1 873 119

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr1121

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P371588 NM_146348.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P371588 NM_146348.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P114593 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P371588 NM_146348.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P371588 NM_146348.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P371588 NM_146348.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P371588 NM_146348.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21405721 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21405718 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21405709 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21405699 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21405691 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21405686 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21405712 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21405720 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21405694 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21405711 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21405680 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21405688 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21405683 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21405704 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21405705 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21405719 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21405698 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21405701 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21405703 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21405707 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21405717 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21405682 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21405695 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21405700 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21405713 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21405681 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21405696 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21405689 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21405702 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21405692 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P114593 - -
          Whole Mouse Genome G4122A A_51_P371588 NM_146348.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P371588 NM_146348.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P100132 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P114593 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01909335 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01909327 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01909317 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02016083 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21405728 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21405705 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21405689 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21405709 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21405695 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21405696 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21405688 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21405727 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21405707 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21405730 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21405742 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21405702 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21405682 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21405711 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21405740 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21405717 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21405725 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21405680 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21405731 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21405677 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21405698 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21405691 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21405713 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21405686 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21405719 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21405700 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21405699 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21405721 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21405703 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21405692 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21405694 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21405701 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21405704 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21405720 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405707 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405695 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405682 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405700 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405713 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02266515 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405725 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405699 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405709 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405705 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405701 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405733 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405683 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405696 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405727 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02021608 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405723 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405691 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405692 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405704 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405717 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405684 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405677 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405681 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405719 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405720 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405689 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405711 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01830286 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405674 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405702 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405712 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405698 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405675 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405721 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405688 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405686 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405703 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02100442 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405728 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10473532 - -
          Affymetrix Mouse Gene 1.1 ST Array 10473531 - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 806613 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 1410372 - -
          Illumina mouse-6 v1.0 expression beadchip 1410372 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001410372 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1248317 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1248317 - -
          mouse-6 v1.1 (Illumina) scl4916.1.1_229-S - -
          Illumina Sentrix mouse-6.v1 bead array 1410372 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl4916.1.1_229-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1410372 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1248317 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1248317 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-4lm45:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0