Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00526303_s1
          See other OLFR633 GE Assays ›
          Gene Symbol:
          Olfr633
          Gene Name:
          olfactory receptor 633
          Gene Aliases:
          MOR12-5
          Chromosome Location:
          Chr.7: 103946568 - 103947506 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00526303_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr633
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146354.1 NP_666466.1 1 51 69

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700001511.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr633

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P240632 NM_146354.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P240632 NM_146354.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P108752 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P240632 NM_146354.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P240632 NM_146354.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P240632 NM_146354.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P240632 NM_146354.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25385803 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25385810 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25385806 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25385805 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25385804 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25385802 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25385808 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P108752 - -
          Whole Mouse Genome G4122A A_52_P240632 NM_146354.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P240632 NM_146354.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P107967 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P122272 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P108752 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25385806 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25385894 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25385821 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25385861 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25385824 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25385862 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25385798 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25385842 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25385907 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25385902 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25385846 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25385883 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25385863 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25385923 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25385813 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25385817 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25385802 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25385869 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25385810 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25385819 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25385867 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25385828 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25385791 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25385800 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25385899 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25385826 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25385890 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25385839 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25385796 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25385914 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25385845 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25385836 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25385792 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25385852 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25385805 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25385873 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25385841 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25385803 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25385909 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25385850 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25385879 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25385834 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25385816 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25385918 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25385912 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25385917 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25385888 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25385911 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25385831 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25385808 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25385904 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25385885 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25385921 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25385920 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25385794 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25385843 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25385823 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25385830 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25385790 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25385881 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25385837 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25385919 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25385866 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25385799 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25385854 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25385833 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25385897 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25385871 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25385895 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25385875 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25385847 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25385801 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25385818 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25385859 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25385877 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25385865 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25385850 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25385859 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25385810 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25385833 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25385843 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25385917 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25385912 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05945213 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25385854 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25385805 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25385804 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25385871 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25385818 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25385890 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25385831 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25385835 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25385866 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25385846 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25385819 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25385883 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06210059 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25385869 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25385816 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25385830 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25385877 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06303800 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25385823 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06417030 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25385914 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25385841 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06303804 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25385842 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25385814 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25385863 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25385875 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25385867 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25385862 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25385813 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25385921 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25385918 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25385838 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25385879 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25385897 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25385919 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25385894 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25385907 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25385803 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25385808 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25385925 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06303786 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25385836 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25385826 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25385885 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25385839 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25385817 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25385827 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25385845 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25385828 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25385895 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25385861 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25385874 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25385806 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25385865 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25385847 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25385855 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25385881 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25385837 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25385821 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25385911 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25385909 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25385852 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25385884 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25385888 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10555835 - -
          Affymetrix Mouse Gene 1.1 ST Array 10555834 - -
          Affymetrix Mouse Clariom S Array PSR0700012806.mm.1 TC0700001511.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700012806.mm.1 NM_146354.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700012806.mm.1 NM_146354.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 541439 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6510041 - -
          Illumina mouse-6 v1.0 expression beadchip 6510041 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006510041 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1222439 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1222439 - -
          mouse-6 v1.1 (Illumina) scl0258351.2_187-S - -
          Illumina Sentrix mouse-6.v1 bead array 6510041 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0258351.2_187-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6510041 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1222439 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1222439 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0258351.2_187-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-jwmpg:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0