Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00526342_s1
          See other OLFR677 GE Assays ›
          Gene Symbol:
          Olfr677
          Gene Name:
          olfactory receptor 677
          Gene Aliases:
          MOR32-11
          Chromosome Location:
          Chr.7: 105056248 - 105057186 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00526342_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr677
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146358.1 NP_666470.1 1 58 86

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700001541.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr677

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P211351 NM_146358.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P211351 NM_146358.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P173112 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P211351 NM_146358.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P211351 NM_146358.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P211351 NM_146358.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P211351 NM_146358.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P173112 - -
          Whole Mouse Genome G4122A A_51_P211351 NM_146358.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P211351 NM_146358.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P113316 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P102568 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P173112 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25390495 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25390589 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25390540 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25390523 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25390591 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25390487 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25390532 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25390516 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25390544 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25390507 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25390547 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25390527 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25390587 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25390555 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25390511 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25390551 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25390522 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25390503 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25390502 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25390593 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25390517 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25390499 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25390483 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25390592 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25390497 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25390482 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25390549 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25390557 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25390506 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25390504 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25390518 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25390514 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25390552 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25390531 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25390534 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25390539 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25390537 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25390536 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25390530 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25390494 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25390488 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25390521 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25390498 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25390525 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25390597 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25390513 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25390600 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25390490 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25390541 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25390506 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06515485 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25390557 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25390551 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25390498 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25390531 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25390511 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25390534 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25390490 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25390533 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25390532 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25390522 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25390530 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25390517 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25390497 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25390520 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25390488 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25390546 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25390523 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25390521 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25390525 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25390549 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25390495 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25390516 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25390513 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25390514 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25390507 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25390504 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25390491 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25390541 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25390536 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25390552 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25390539 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25390518 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06061053 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25390494 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25390540 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25390499 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25390547 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25390500 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25390502 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25390544 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25390555 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25390487 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10555906 - -
          Affymetrix Mouse Gene 1.1 ST Array 10555905 - -
          Affymetrix Mouse Clariom S Array PSR0700012931.mm.1 TC0700001541.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700012931.mm.1 NM_146358.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700012931.mm.1 NM_146358.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 859839 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 520040 - -
          Illumina mouse-6 v1.0 expression beadchip 520040 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000520040 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1251653 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1251653 - -
          mouse-6 v1.1 (Illumina) scl32246.1.1_88-S - -
          Illumina Sentrix mouse-6.v1 bead array 520040 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl32246.1.1_88-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 520040 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1251653 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1251653 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0