Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00526410_s1
          See other OLFR1094 GE Assays ›
          Gene Symbol:
          Olfr1094
          Gene Name:
          olfactory receptor 1094
          Gene Aliases:
          MOR179-7
          Chromosome Location:
          Chr.2: 86828676 - 86829822 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00526410_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr1094
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146365.2 NP_666477.1 1 334 143
          GenBank mRNA BC120867.1 - 1 334 40129790 143

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200001274.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr1094

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          signal transduction
          G-protein coupled receptor signaling pathway
          sensory perception of smell
          response to stimulus
          detection of chemical stimulus involved in sensory perception of smell
          signal transducer activity
          G-protein coupled receptor activity
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2178660 NM_146365.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2178660 BC120867.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P163550 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2178660 NM_146365.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2178660 BC120867.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P442736 NM_146365.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21403772 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21403789 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21403793 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21403782 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21403760 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21403756 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21403776 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21403750 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21403777 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21403791 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21403754 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21403758 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21403767 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21403749 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21403762 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21403770 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21403743 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21403778 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21403745 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21403741 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21403746 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21403740 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21403779 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21403748 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21403766 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21403755 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21403752 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21403757 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21403788 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21403764 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21403763 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21403774 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21403795 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21403751 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21403784 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21403781 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P163550 - -
          Whole Mouse Genome G4122A A_51_P442736 NM_146365.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P104689 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P163550 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01292333 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01292322 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01292320 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01260231 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01292332 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21403752 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21403778 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21403784 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21403744 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21403793 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21403758 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21403781 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21403770 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21403788 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21403750 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21403763 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21403777 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21403774 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21403800 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21403762 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21403760 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21403782 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21403776 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21403772 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21403746 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21403792 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21403755 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21403767 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21403745 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21403805 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21403754 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21403756 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21403762 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21403776 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21403741 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21403772 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21403774 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21403800 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21403767 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01296592 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21403782 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01419249 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21403791 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21403778 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21403748 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21403784 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21403750 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21403760 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21403797 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21403746 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21403777 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21403756 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21403770 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21403745 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21403755 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21403740 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21403763 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21403798 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21403792 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21403795 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21403803 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21403793 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21403781 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21403754 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21403788 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21403758 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21403752 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21403744 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10473510 - -
          Affymetrix Mouse Gene 1.1 ST Array 10473509 - -
          Affymetrix Mouse Clariom S Array PSR0200010010.mm.1 NM_146365.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200010010.mm.1 NM_146365.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200010010.mm.1 TC0200001274.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 763743 - -
          ABI Mouse Genome Survey Microarray 774471 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4230403 - -
          Illumina mouse-6 v1.0 expression beadchip 4230403 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004230403 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2773408 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2773408 - -
          mouse-6 v1.1 (Illumina) scl4924.1.1_49-S - -
          Illumina Sentrix mouse-6.v1 bead array 4230403 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl4924.1.1_49-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4230403 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2957580 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2773408 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2957580 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-jwmpg:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0