Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00526446_s1
          See other OLFR361 GE Assays ›
          Gene Symbol:
          Olfr361
          Gene Name:
          olfactory receptor 361
          Gene Aliases:
          MOR159-3
          Chromosome Location:
          Chr.2: 37084778 - 37085746 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00526446_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr361
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146368.1 NP_666480.1 1 924 85
          GenBank mRNA BC104114.1 - 1 922 85
          BC104115.1 - 1 922 85
          BC114393.1 - 1 923 85
          BC114424.1 - 1 923 85

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr361

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P364871 BC114424.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P364871 NM_146368.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P364871 BC104114.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P364871 BC104115.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P364871 BC114393.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P364871 BC114424.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P364871 NM_146368.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P364871 BC104114.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P364871 BC104115.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P364871 BC114393.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P124785 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P364871 BC114424.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P364871 NM_146368.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P364871 BC104114.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P364871 BC104115.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P364871 BC114393.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P364871 BC114424.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P364871 NM_146368.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P364871 BC104114.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P364871 BC104115.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P364871 BC114393.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P364871 BC114424.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P364871 NM_146368.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P364871 BC104114.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P364871 BC104115.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P364871 BC114393.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P364871 BC114424.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P364871 NM_146368.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P364871 BC104114.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P364871 BC104115.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P364871 BC114393.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P124785 - -
          Whole Mouse Genome G4122A A_51_P364871 BC114424.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P364871 NM_146368.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P364871 BC104114.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P364871 BC104115.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P364871 BC114393.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P364871 BC114424.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P364871 NM_146368.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P364871 BC104114.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P364871 BC104115.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P364871 BC114393.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P109309 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P124785 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21140894 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21140867 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21140871 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21140917 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21140915 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21140837 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21140909 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21140921 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21140888 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21140904 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21140922 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21140847 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21140914 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21140878 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21140891 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21140902 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21140924 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21140919 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21140903 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21140889 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21140839 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21140842 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21140897 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21140876 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21140887 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21140901 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21140881 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21140883 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21140884 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21140929 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21140908 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21140895 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21140854 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21140879 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21140890 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21140893 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21140861 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21140840 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21140878 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21140909 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21140899 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21140842 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01987273 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01987275 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02172958 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21140879 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01987276 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01895664 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21140904 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01627048 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21140876 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21140860 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21140852 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21140908 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21140902 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21140849 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21140861 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01987272 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21140901 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21140881 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01987285 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21140891 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21140867 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01627074 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21140915 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21140859 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02172967 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21140858 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21140914 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21140903 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21140897 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02172970 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21140839 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21140890 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21140889 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21140888 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21140864 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21140905 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21140912 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21140885 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21140883 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21140910 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10482134 - -
          Affymetrix Mouse Gene 1.1 ST Array 10482133 - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 790428 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 460019 - -
          Illumina mouse-6 v1.0 expression beadchip 460019 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000460019 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2649744 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2649744 - -
          mouse-6 v1.1 (Illumina) scl4521.1.1_121-S - -
          Illumina Sentrix mouse-6.v1 bead array 460019 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl4521.1.1_121-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 460019 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1225711 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1225711 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline