Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00526651_s1
          See other OLFR771 GE Assays ›
          Gene Symbol:
          Olfr771
          Gene Name:
          olfactory receptor 771
          Gene Aliases:
          MOR114-8
          Chromosome Location:
          Chr.10: 129160029 - 129160982 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00526651_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr771
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146547.1 NP_666758.1 1 444 65

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1000003177.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr771

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2057751 NM_146547.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P170830 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P145591 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P104567 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2057751 NM_146547.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P479138 NM_146547.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27544933 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27544944 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27544943 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27544968 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27544941 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27544954 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27544932 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27544964 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27544960 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27544940 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27544946 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27544956 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27544934 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27544947 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27544953 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27544951 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27544945 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27544965 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27544962 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27544927 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27544923 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27544931 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27544938 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27544958 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27544966 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27544936 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27544949 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27544929 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27544928 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27544930 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P145591 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P104567 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P170830 - -
          Whole Mouse Genome G4122A A_51_P479138 NM_146547.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P127433 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P170830 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P104567 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P145591 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08943382 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08943391 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08943390 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08943378 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08943385 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08943396 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08943397 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27544949 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27544954 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27544920 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27544907 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27544964 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27544938 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27544977 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27544979 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27544930 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27544915 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27544904 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27544956 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27544981 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27544894 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27544899 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27544943 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27544940 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27544917 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27544933 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27544923 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27544962 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27544905 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27544960 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27544966 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27544934 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27544909 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27544919 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27544951 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27544936 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27544958 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27544928 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27544967 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27544932 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27544983 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27544973 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27544945 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27544901 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27544918 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27544931 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27544896 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27544900 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27544974 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27544943 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27544933 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27544947 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08724959 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27544931 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27544966 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27544915 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27544907 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27544928 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27544909 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27544940 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27544960 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27544919 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08569920 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27544918 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27544911 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27544938 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27544954 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27544906 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27544969 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27544973 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27544945 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27544977 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27544970 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27544964 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27544936 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27544962 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27544971 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08425310 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27544917 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27544958 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27544905 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27544951 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08926787 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27544932 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27544920 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27544967 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27544934 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27544956 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27544921 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08946670 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27544981 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27544930 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08943387 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08926788 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27544923 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10373619 - -
          Affymetrix Mouse Gene 1.1 ST Array 10373618 - -
          Affymetrix Mouse Clariom S Array PSR1000023947.mm.1 NM_146547.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000023947.mm.1 NM_146547.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000023947.mm.1 TC1000003177.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 810680 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 3450064 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003450064 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2697776 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2697776 - -
          mouse-6 v1.1 (Illumina) scl5164.1.1_48-S - -
          Illumina Sentrix mouse-6.v1 bead array 3450064 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl5164.1.1_48-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3450064 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2901331 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2901331 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-jwmpg:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0