Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00526844_s1
          See other OLFR1043 GE Assays ›
          Gene Symbol:
          Olfr1043
          Gene Name:
          olfactory receptor 1043
          Gene Aliases:
          MOR185-7
          Chromosome Location:
          Chr.2: 86162003 - 86162947 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00526844_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr1043
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146577.2 NP_666788.1 1 158 116

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200003917.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr1043

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          G-protein coupled receptor activity
          olfactory receptor activity
          odorant binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P133911 NM_146577.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P133911 NM_146577.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P138515 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P133911 NM_146577.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P133911 NM_146577.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P133911 NM_146577.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P133911 NM_146577.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P138515 - -
          Whole Mouse Genome G4122A A_51_P133911 NM_146577.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P133911 NM_146577.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P125022 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P138515 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401031 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401028 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400951 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400928 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400943 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400955 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401037 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400945 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400959 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401011 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401030 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400998 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400977 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400947 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400990 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400922 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400949 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401026 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401035 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401006 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401047 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400996 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400964 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400953 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401024 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401029 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400956 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400975 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400923 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400992 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400961 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401032 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401021 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400972 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401019 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401001 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401003 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400986 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401016 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400970 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400994 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401041 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400973 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400988 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400962 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401040 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401004 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401036 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401044 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400971 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401009 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401017 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400958 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400948 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400999 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400969 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401023 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401011 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21400986 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401001 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21400996 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401030 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401032 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21400992 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401024 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401012 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401019 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401021 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21400998 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401020 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401033 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401009 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401026 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401000 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401016 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01783553 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401025 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401013 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401029 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401023 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21400999 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401003 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21400990 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401036 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401034 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01674934 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401031 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401006 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401028 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02092539 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21400988 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401035 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10484564 - -
          Affymetrix Mouse Gene 1.1 ST Array 10484563 - -
          Affymetrix Mouse Clariom S Array PSR0200033733.mm.1 NM_146577.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200033733.mm.1 NM_146577.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200033733.mm.1 TC0200003917.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 424834 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_3161873 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_3161873 - -
          mouse-6 v1.1 (Illumina) GI_22129490-S - -
          MouseRef-8 v2 Expression BeadChip ILMN_3161873 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3161873 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline