Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00526890_s1
          See other OLFR1046 GE Assays ›
          Gene Symbol:
          Olfr1046
          Gene Name:
          olfactory receptor 1046
          Gene Aliases:
          MOR194-1
          Chromosome Location:
          Chr.2: 86216758 - 86217708 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00526890_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr1046
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146582.2 NP_666793.2 1 278 91
          GenBank mRNA BC099891.1 - 1 277 91
          BC099892.1 - 1 277 91
          BC103540.1 - 1 277 91

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr1046

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          G-protein coupled receptor activity
          olfactory receptor activity
          odorant binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1981469 BC099892.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1981469 BC103540.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1981469 NM_146582.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1981469 BC099891.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P114847 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1981469 BC099892.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1981469 BC103540.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1981469 NM_146582.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1981469 BC099891.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P431684 BC099892.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P431684 BC103540.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P431684 NM_146582.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P431684 BC099891.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P114847 - -
          Whole Mouse Genome G4122A A_51_P431684 BC099892.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P431684 BC103540.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P431684 NM_146582.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P431684 BC099891.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P100089 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P114847 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401235 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401308 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401328 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401335 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401313 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401263 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401312 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401302 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401337 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401332 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401264 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401265 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401329 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401243 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401304 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401330 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401324 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401331 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01641102 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01641124 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401302 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401260 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401335 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401314 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01317590 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01979302 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401265 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401332 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401253 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401304 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401326 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01586827 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02080786 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401330 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401329 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401308 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401311 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01451174 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401339 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401267 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01586707 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401320 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401300 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401317 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01641101 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401321 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401331 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401312 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401337 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401262 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01962917 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401259 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01774036 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01586706 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401263 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10484572 - -
          Affymetrix Mouse Gene 1.1 ST Array 10484571 - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 753482 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 2190458 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002190458 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2636238 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2636238 - -
          mouse-6 v1.1 (Illumina) scl0258575.1_253-S - -
          Illumina Sentrix mouse-6.v1 bead array 2190458 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0258575.1_253-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2190458 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2636238 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2636238 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline