Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00526909_s1
          See other OLFR1018 GE Assays ›
          Gene Symbol:
          Olfr1018
          Gene Name:
          olfactory receptor 1018
          Gene Aliases:
          MOR260-5
          Chromosome Location:
          Chr.2: 85822973 - 85823908 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00526909_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr1018
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146586.2 NP_666797.2 1 247 66

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200001250.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor Detect Genomic DNA Recommended Assay Mus musculus Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr1018

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1954226 NM_146586.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P116868 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1954226 NM_146586.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P420519 NM_146586.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21399498 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21399499 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21399502 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21399501 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21399536 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21399532 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21399537 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21399511 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21399526 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21399525 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21399512 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21399508 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21399488 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21399494 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21399519 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21399538 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21399496 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21399503 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21399528 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21399524 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21399493 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21399486 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21399521 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21399517 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21399492 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21399541 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21399505 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21399506 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21399531 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21399491 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21399535 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21399523 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21399514 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P116868 - -
          Whole Mouse Genome G4122A A_51_P420519 NM_146586.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P116868 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01568304 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01449885 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01774543 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01774542 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01568310 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21399525 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21399517 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21399528 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21399476 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21399482 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21399546 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21399477 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21399484 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21399506 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21399553 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21399521 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21399488 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21399508 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21399496 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21399535 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21399502 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21399511 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21399491 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21399494 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21399512 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21399503 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21399526 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21399519 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21399549 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21399537 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21399498 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21399481 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21399514 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21399501 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21399547 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21399493 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21399536 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21399538 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21399479 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21399532 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21399529 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21399474 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21399531 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21399545 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21399523 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21399479 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01449889 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21399528 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21399555 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21399484 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21399505 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21399553 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21399538 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21399512 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21399524 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21399547 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21399481 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21399545 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21399536 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02190916 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21399526 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21399506 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21399500 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21399491 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21399535 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21399488 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21399543 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21399511 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21399501 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02246287 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21399474 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21399514 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21399494 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21399502 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21399529 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21399521 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21399517 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21399493 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21399496 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21399477 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21399549 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21399531 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21399486 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21399503 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21399519 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21399546 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21399508 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21399537 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21399476 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21399541 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21399532 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01332852 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10473468 - -
          Affymetrix Mouse Gene 1.1 ST Array 10473467 - -
          Affymetrix Mouse Clariom S Array PSR0200009958.mm.1 NM_146586.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200009958.mm.1 NM_146586.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200009958.mm.1 TC0200001250.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 1660400 - -
          Illumina mouse-6 v1.0 expression beadchip 1660400 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001660400 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1232618 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1232618 - -
          mouse-6 v1.1 (Illumina) scl4940.1.1_266-S - -
          Illumina Sentrix mouse-6.v1 bead array 1660400 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl4940.1.1_266-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1660400 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1232618 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1232618 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-mrft7:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0