Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00526956_s1
          See other OLFR1101 GE Assays ›
          Gene Symbol:
          Olfr1101
          Gene Name:
          olfactory receptor 1101
          Gene Aliases:
          MOR179-3
          Chromosome Location:
          Chr.2: 86988242 - 86989174 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00526956_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr1101
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146591.2 NP_666802.2 1 454 88

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr1101

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          cell-cell signaling
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          adenylate cyclase-activating adrenergic receptor signaling pathway
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2082713 NM_146591.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P154000 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2082713 NM_146591.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P412895 NM_146591.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21404354 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21404361 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21404347 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21404358 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21404351 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21404363 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21404352 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21404349 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21404353 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21404345 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21404348 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21404357 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21404364 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21404360 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P154000 - -
          Whole Mouse Genome G4122A A_51_P412895 NM_146591.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P134903 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P154000 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404364 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404363 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404347 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404279 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404311 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404282 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404351 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404293 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404319 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404295 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404336 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404318 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404327 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404284 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404352 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404353 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404291 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404316 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404344 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404271 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404288 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404358 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404333 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404322 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404286 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404312 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404329 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404360 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404323 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404287 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404270 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404281 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404337 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404339 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404349 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404357 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404275 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404274 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404277 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404334 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404361 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404313 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404354 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404290 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404326 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404337 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01896598 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404313 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404329 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404334 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404317 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404333 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404339 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01544299 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404312 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404319 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404311 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404316 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01896593 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404323 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404322 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404325 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01806588 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404327 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10484645 - -
          Affymetrix Mouse Gene 1.1 ST Array 10484644 - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 642167 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4810020 - -
          Illumina mouse-6 v1.0 expression beadchip 4810020 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004810020 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2772747 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2772747 - -
          mouse-6 v1.1 (Illumina) scl4323.1.1_95-S - -
          Illumina Sentrix mouse-6.v1 bead array 4810020 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl4323.1.1_95-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4810020 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2772747 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2772747 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline