Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00526980_s1
          See other OLFR1100 GE Assays ›
          Gene Symbol:
          Olfr1100
          Gene Name:
          olfactory receptor 1100
          Gene Aliases:
          MOR206-4
          Chromosome Location:
          Chr.2: 86977829 - 86978794 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00526980_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr1100
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146594.1 NP_666805.1 1 256 119
          GenBank mRNA BC130249.1 - 1 260 40142420 119
          BC139802.1 - 1 260 40142396 119

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200003963.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr1100

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          G-protein coupled receptor activity
          olfactory receptor activity
          odorant binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P350942 NM_146594.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P350942 BC130249.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P350942 BC139802.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P350942 NM_146594.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P350942 BC130249.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P350942 BC139802.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 187169 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P109123 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P350942 NM_146594.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P350942 BC130249.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P350942 BC139802.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P350942 NM_146594.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P350942 BC130249.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P350942 BC139802.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P350942 NM_146594.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P350942 BC130249.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P350942 BC139802.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P350942 NM_146594.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P350942 BC130249.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P350942 BC139802.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P109123 - -
          Whole Mouse Genome G4122A A_51_P350942 NM_146594.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P350942 BC130249.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P350942 BC139802.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P350942 NM_146594.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P350942 BC130249.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P350942 BC139802.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P132610 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P125897 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P109123 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404256 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404245 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404259 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404281 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404311 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404274 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404305 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404312 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404279 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404254 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404267 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404287 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404284 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404271 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404261 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404258 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404295 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404300 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404282 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404291 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404263 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404262 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404307 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404270 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404293 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404275 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404246 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404313 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404303 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404288 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404290 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404286 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404277 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404274 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404290 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404267 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01934350 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404303 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404261 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404300 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404254 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404307 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404281 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404295 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404258 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404282 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01934356 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404256 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01173365 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404252 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404291 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404305 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404288 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404259 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404266 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404293 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404277 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404279 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404271 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404275 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404297 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404270 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404265 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404262 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404263 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10484643 - -
          Affymetrix Mouse Gene 1.1 ST Array 10484642 - -
          Affymetrix Mouse Clariom S Array PSR0200033826.mm.1 NM_146594.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200033826.mm.1 NM_146594.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200033826.mm.1 TC0200003963.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 806276 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 1450451 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001450451 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1242841 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1242841 - -
          mouse-6 v1.1 (Illumina) scl4324.1.1_195-S - -
          Illumina Sentrix mouse-6.v1 bead array 1450451 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl4324.1.1_195-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1450451 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1242841 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1242841 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline